Skip to Content
Merck
All Photos(2)

Key Documents

112429

Sigma-Aldrich

2-Ethylpyridine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H9N
CAS Number:
Molecular Weight:
107.15
Beilstein:
106480
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.496 (lit.)

bp

149 °C (lit.)

density

0.937 g/mL at 25 °C (lit.)

SMILES string

CCc1ccccn1

InChI

1S/C7H9N/c1-2-7-5-3-4-6-8-7/h3-6H,2H2,1H3

InChI key

NRGGMCIBEHEAIL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Ethylpyridine linked with silica is a popular stationary phase for chiral and achiral separations in supercritical fluid chromatography.

Application

2-Ethylpyridine is used to study its effect on the proliferation and survival of human umbilical vein endothelial cells (HUVECs), HMVECs from lung, and NIH 3T3 cells.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

102.2 °F - closed cup

Flash Point(C)

39 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

S Muthu et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 93, 214-222 (2012-04-07)
Fourier transform Raman and Fourier transform infrared spectra of 2-ethylpyridine-4-carbothioamide were recorded in the regions 3600-100 cm(-1) and 4000-450 cm(-1), respectively in the solid phase. 2-Ethylpyridine-4-carbothioamide is used as anti-tubercular agent that inhibits mycolic acid synthesis. The equilibrium geometry harmonic
E J O'Loughlin et al.
Biodegradation, 10(2), 93-104 (1999-08-31)
A bacterium capable of degrading 2-methylpyridine was isolated by enrichment techniques from subsurface sediments collected from an aquifer located at an industrial site that had been contaminated with pyridine and pyridine derivatives. The isolate, identified as an Arthrobacter sp., was
David Speybrouck et al.
Current topics in medicinal chemistry, 12(11), 1250-1263 (2012-05-11)
Supercritical fluid (SF) was discovered 200 years ago, but the use of this fluid as a mobile phase in chromatography only became popular fifty years ago. The development of the supercritical fluid chromatography (SFC) was progressing slowly due to technological
Richard Yu et al.
Toxicological sciences : an official journal of the Society of Toxicology, 93(1), 82-95 (2006-06-23)
Our purpose was to determine the effects of six cigarette toxicants (pyridine, nicotine, 2-ethylpyridine, 3-ethylpyridine, p-cresol, and pyrazine) on three types of cultured mammalian cells (human umbilical vein endothelial cells [HUVECs], human microvascular endothelial cells [HMVECs], and NIH 3T3 cells)
Cleidiane G Zampronio et al.
The Analyst, 127(8), 1054-1060 (2002-08-28)
Direct sampling tandem mass spectrometry (MS/MS) was used for the quantitation of mixtures of the isomers 2-, 3- and 4-ethyl pyridine. The similarity between the analytes and the second-order nature of MS/MS data require the use of multivariate calibration techniques

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service