Skip to Content
Merck
All Photos(2)

Key Documents

C95005

Sigma-Aldrich

4-Pyridinecarbonitrile

98%

Synonym(s):

4-Cyanopyridine, Isonicotinic acid nitrile, Isonicotinonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4N2
CAS Number:
Molecular Weight:
104.11
Beilstein:
107712
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

98%

form

crystals

mp

76-79 °C (lit.)

SMILES string

N#Cc1ccncc1

InChI

1S/C6H4N2/c7-5-6-1-3-8-4-2-6/h1-4H

InChI key

GPHQHTOMRSGBNZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

190.4 °F

Flash Point(C)

88 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Chao Hong et al.
Pharmaceutical research, 32(1), 47-60 (2014-06-19)
To develop a streamlined strategy for pharmaceutical cocrystal preparation without knowledge of the stoichiometric ratio by preparing and characterizing the cocrystals of myricetin (MYR) with four cocrystal coformers (CCF). An approach based on the phase solubility diagram (PSD) was used
Masazumi Tamura et al.
Nature communications, 6, 8580-8580 (2015-10-06)
Multidentate materials formed by simply mixing heterogeneous and homogeneous components are promising for construction of versatile active sites on the surface of heterogeneous compounds, however, to the best of our knowledge, there are no reports on such materials. Self-assembly of

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service