Merck
  • cIEF for rapid pKa determination of small molecules: a proof of concept.
cIEF for rapid pKa determination of small molecules: a proof of concept.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences (2014-07-06)
Stéphanie Romand, Julie Schappler, Jean-Luc Veuthey, Pierre-Alain Carrupt, Sophie Martel
ABSTRACT
A capillary isoelectric focusing (cIEF) method was developed for the determination of the ionization constants (pKa) of small molecules. Two approaches used to decrease the electroosmotic flow (EOF) were compared: (i) a hydroxypropylcellulose (HPC) coated capillary in aqueous medium and (ii) the addition of glycerol to act as a viscosifying agent. The cIEF method with the glycerol medium was selected, and the ionization constants of 22 basic and 21 acidic compounds, including 15 pharmaceutical drugs, were determined, resulting in pKa values from 3.5 to 7.4 and 6.4 to 9.3, respectively. cIEF offers an interesting alternative to other techniques for pKa determination with low sample consumption, high throughput and low cost.
Sigma-Aldrich
4-Nitrophenol solution, 10 mM
Sigma-Aldrich
2,4-Dibromophenol, 95%
Supelco
Nitrofurantoin, VETRANAL®, analytical standard
Supelco
Aniline, analytical standard
Sigma-Aldrich
3-Fluorophenol, 98%
Sigma-Aldrich
Aniline, ACS reagent, ≥99.5%
Sigma-Aldrich
Trimethoprim, ≥99.0% (HPLC)
Supelco
Trimethoprim, VETRANAL®, analytical standard
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Isoquinoline, 97%
Sigma-Aldrich
4-Methylpyridine, 98%
Sigma-Aldrich
4-Methylpyridine, 99%
Sigma-Aldrich
Aniline, ReagentPlus®, 99%
Sigma-Aldrich
p-Anisidine, 99%
Sigma-Aldrich
2-Nitrophenol, 98%
Sigma-Aldrich
4-Nitrophenol, ReagentPlus®, ≥99%
Sigma-Aldrich
2,4-Dichlorophenol, 99%
Sigma-Aldrich
4-Chloroaniline, purified by sublimation, ≥99%
Sigma-Aldrich
Methyl nicotinate, ≥99%, FG
Sigma-Aldrich
2,6-Dibromophenol, 99%
Sigma-Aldrich
N,N-Diethylaniline, ≥99%
Sigma-Aldrich
2,6-Dichlorophenol, 99%
Sigma-Aldrich
2,4-Dimethylaniline, ≥99%
Sigma-Aldrich
4-Nitrophenol, spectrophotometric grade
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Sigma-Aldrich
Fluorouracil, meets USP testing specifications
Supelco
4-Chloroaniline, PESTANAL®, analytical standard
Sigma-Aldrich
Methyl nicotinate, puriss., ≥99.0% (GC)
Sigma-Aldrich
Trimethoprim, ≥98.5%
Sigma-Aldrich
4-Chloroaniline, 98%