Skip to Content
Merck
All Photos(3)

Key Documents

M6182

Sigma-Aldrich

Mercaptosuccinic acid

97%

Synonym(s):

Thiomalic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOCCH(SH)CH2COOH
CAS Number:
Molecular Weight:
150.15
Beilstein:
1099858
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

mp

155-157 °C (lit.)

SMILES string

OC(=O)CC(S)C(O)=O

InChI

1S/C4H6O4S/c5-3(6)1-2(9)4(7)8/h2,9H,1H2,(H,5,6)(H,7,8)

InChI key

NJRXVEJTAYWCQJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Mercaptosuccinic acid(MSA) is a chiral multifunctional intermediate material used as a stabilizing and capping agent in the synthesis of biologically active sulfurs.

Application

MSA has been used to cap cadmium telluride quantum dots(QDs) and nanocrystals for a wide range of applications like detection of benzoic acid and total paraben based preservatives in food items, biological fluorescent labelling in bio-imaging and can bond with antibodies or antigens for biomedical applications. MSA may be coated on gold nanoparticles(AuNPs) as a reducing agent and a surface modifier that finds potential application in fluorescent detection of mercury.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The solubility of mercaptosuccinic acid in water+ (methanol, ethanol, acetone) mixtures from (278.15 to 333.15 K).
Kai Y, et al.
Fluid Phase Equilibria, 361, 282-288 (2014)
Successive detection of benzoic acid and total parabens in foodstuffs using mercaptosuccinic acid capped cadmium telluride quantum dots.
Prapainop K, et al.
Food Control, 96, 508-516 (2018)
Mercaptosuccinic acid-coated NIR-emitting gold nanoparticles for the sensitive and selective detection of Hg2+.
Xiong X, et al.
Spectrochimica Acta. Part A, Molecular and Biomolecular Spectroscopy, 188(9), 483-487 (2018)
Siqiu Ye et al.
The Analyst, 137(14), 3365-3371 (2012-06-05)
A colorimetric sensor for Cu(II) ions has been developed based on mixed self-assembled monolayers (SAMs) modified gold nanoparticles (AuNPs). The AuNPs were modified with mixed SAMs consisting of mercaptosuccinic acid and the product of electrochemically triggered Michael addition reaction of
Ming Yan et al.
Toxicology, 282(3), 94-103 (2011-02-05)
Quantum dots (QDs), as novel bioimaging and drug delivery agents, are generally introduced into vascular system by injection, and thus directly exposed to vascular endothelial cells (ECs). However, the adverse effects of QDs on ECs are poorly understood. In this

Articles

Solvothermal synthesis of nanoparticles: applications from nanocircuits and nano-optical circuits to nanomagnetics and biotech.

Solvothermal synthesis of nanoparticles: applications from nanocircuits and nano-optical circuits to nanomagnetics and biotech.

Solvothermal synthesis of nanoparticles: applications from nanocircuits and nano-optical circuits to nanomagnetics and biotech.

Solvothermal synthesis of nanoparticles: applications from nanocircuits and nano-optical circuits to nanomagnetics and biotech.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service