Merck
  • Enhanced photodynamic therapy efficacy of methylene blue-loaded calcium phosphate nanoparticles.
Enhanced photodynamic therapy efficacy of methylene blue-loaded calcium phosphate nanoparticles.
Journal of photochemistry and photobiology. B, Biology (2015-03-22)
Da-Young Seong, Young-Jin Kim
ABSTRACT
Although methylene blue (MB) is the most inexpensive photosensitizer with promising applications in the photodynamic therapy (PDT) for its high quantum yield of singlet oxygen generation, the clinical use of MB has been limited by its rapid enzymatic reduction in the biological environment. To enhance PDT efficacy of MB by preventing the enzymatic reduction, we have developed a new mineralization method to produce highly biocompatible MB-loaded calcium phosphate (CaP-MB) nanoparticles in the presence of polymer templates. The resulting CaP-MB nanoparticles exhibited spherical shape with a size of under 50 nm. Fourier transform infrared (FT-IR) and zeta-potential analyses confirmed the insertion of MB into the CaP-MB nanoparticles. The encapsulation of MB in CaP nanoparticles could effectively protect MB from the enzymatic reduction. In addition, the CaP-MB nanoparticles exhibited a good biocompatibility in the dark condition and significantly enhanced PDT efficacy due to apoptotic cell death against human breast cancer cells as compared with free MB, implying that CaP-MB nanoparticle system might be potentially applicable in PDT.
Sigma-Aldrich
Adenine, BioReagent, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Adenine, ≥99%
Sigma-Aldrich
Ethylene glycol, JIS special grade, ≥99.5%
Sigma-Aldrich
Ethylene glycol, SAJ first grade, ≥99.0%
Sigma-Aldrich
Adenine, BioReagent, suitable for cell culture
Supelco
Ethylene glycol solution, NMR reference standard, 80% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Ethylene glycol, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Ethylene glycol, anhydrous, 99.8%
Sigma-Aldrich
5-(N,N-Dimethyl)amiloride hydrochloride
Sigma-Aldrich
Methylene blue, certified by the Biological Stain Commission
Sigma-Aldrich
Calcium nitrate tetrahydrate, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Calcium nitrate tetrahydrate, BioXtra, ≥99.0%
Sigma-Aldrich
Methylene Blue Solution 1.4%(w/v)95%ethanol | 7220-79-3, 1.4 % (w/v) in 95% ethanol
Sigma-Aldrich
Methylene Blue solution
Sigma-Aldrich
Ammonium phosphate dibasic, SAJ first grade, 98.5-101.0%
Sigma-Aldrich
Ammonium phosphate dibasic, JIS special grade, ≥99.0%
Sigma-Aldrich
Calcium nitrate tetrahydrate, JIS special grade, ≥98.5%
Sigma-Aldrich
9,10-Dimethylanthracene, 99%
Sigma-Aldrich
Ammonium phosphate dibasic, ≥99.99% trace metals basis
Sigma-Aldrich
Methylene Blue solution, 0.05 wt. % in H2O
Sigma-Aldrich
Ethylene glycol 5 M solution
Sigma-Aldrich
Methylene Blue solution, for microscopy, concentrate according to Ehrlich, concentrated, aqueous solution