Skip to Content
Merck
All Photos(3)

Documents

A5665

Sigma-Aldrich

Adenine

BioReagent, suitable for plant cell culture, ≥99%

Synonym(s):

6-Aminopurine, Vitamin B4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5N5
CAS Number:
Molecular Weight:
135.13
Beilstein:
5777
EC Number:
MDL number:
UNSPSC Code:
10171502
PubChem Substance ID:
NACRES:
NA.72

product line

BioReagent

Quality Level

Assay

≥99%

form

powder

mol wt

Mw 135.13 g/mol

technique(s)

cell culture | plant: suitable

color

white to off-white

mp

>360 °C (lit.)

solubility

0.5 M HCl: soluble 20 mg/mL, clear to slightly hazy, colorless to faintly yellow

application(s)

agriculture

SMILES string

Nc1ncnc2[nH]cnc12

InChI

1S/C5H5N5/c6-4-3-5(9-1-7-3)10-2-8-4/h1-2H,(H3,6,7,8,9,10)

InChI key

GFFGJBXGBJISGV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Adenine (Ade) is a purine base, which is present in nucleic acids. It binds with ribose to form nucleoside adenosine (A) and attach to deoxyribose to form nucleoside deoxyadenosine (dA).

Application

Adenine has been used in yeast extract supplemented (YES) broth and yeast nitrogen base glutamate (YNG)l media.
Adenine may be used as a plant cell culture additive. Adenine Sulphate (25mg/L) in the culture media such as Murashige & Skoog media improves the frequency of multiple shoot production and recovered the chlorotic symptoms of the leaves.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Single-step marker switching in Schizosaccharomyces pombe using a lithium acetate transformation protocol
Brown S D and Lorenz A
Bio-protocol, 6(24) (2016)
Adenine
Encyclopedia of Genetics (2001)
Didier Wion et al.
Nature reviews. Microbiology, 4(3), 183-192 (2006-02-21)
N(6)-methyl-adenine is found in the genomes of bacteria, archaea, protists and fungi. Most bacterial DNA adenine methyltransferases are part of restriction-modification systems. Certain groups of Proteobacteria also harbour solitary DNA adenine methyltransferases that provide signals for DNA-protein interactions. In gamma-proteobacteria
Georg Behrens et al.
AIDS patient care and STDs, 28(4), 168-175 (2014-03-26)
The once daily, single-tablet regimen (STR) combining rilpivirine (RPV), emtricitabine (FTC), and tenofovir disoproxil fumarate (TDF) provides a simplified treatment option for antiretroviral therapy (ART)-naïve patients with baseline HIV-1 RNA (BLVL) of ≤100,000 copies/mL. The aim of this analysis is
Ravindra K Gupta et al.
The Journal of infectious diseases, 207 Suppl 2, S101-S106 (2013-05-25)
Recent data from studies on treatment as prevention (TasP) and preexposure prophylaxis (PrEP) show that antiretroviral drugs can be used in prevention, as well as in treatment. The movement from first-generation antiretroviral therapy (ART) coformulations based on thymidine analogues to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service