Skip to Content
Merck
All Photos(1)

Key Documents

05-0860

Sigma-Aldrich

Calcium nitrate tetrahydrate

JIS special grade, ≥98.5%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ca(NO3)2 · 4H2O
CAS Number:
Molecular Weight:
236.15
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

JIS special grade

Assay

≥98.5%

form

crystalline

availability

available only in Japan

pH

5.5-7.0

mp

44 °C (dec.) (lit.)

SMILES string

O.O.O.O.[Ca++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/Ca.2NO3.4H2O/c;2*2-1(3)4;;;;/h;;;4*1H2/q+2;2*-1;;;;

InChI key

ICSSIKVYVJQJND-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mehdi Razavi et al.
Materials science & engineering. C, Materials for biological applications, 48, 21-27 (2015-01-13)
Although magnesium (Mg) is a unique biodegradable metal which possesses mechanical property similar to that of the natural bone and can be an attractive material to be used as orthopedic implants, its quick corrosion rate restricts its actual clinical applications.
Da-Young Seong et al.
Journal of photochemistry and photobiology. B, Biology, 146, 34-43 (2015-03-22)
Although methylene blue (MB) is the most inexpensive photosensitizer with promising applications in the photodynamic therapy (PDT) for its high quantum yield of singlet oxygen generation, the clinical use of MB has been limited by its rapid enzymatic reduction in
Kalimuthu Pandi et al.
Carbohydrate polymers, 134, 732-739 (2015-10-03)
Adsorption is a significant reaction occurs between adsorbent/water interface for controlling the pollutants in the aqueous environment. In this regard, an eco-magnetic biosorbent was prepared by uniform deposition of magnetic Fe3O4 particles on the surface of nano-hydroxyapatite (n-HAp)/chitosan (CS) nanocomposite
Senthilguru Kulanthaivel et al.
Materials science & engineering. C, Materials for biological applications, 58, 648-658 (2015-10-20)
The present study delineates the synthesis and characterization of cobalt doped proangiogenic-osteogenic hydroxyapatite. Hydroxyapatite samples, doped with varying concentrations of bivalent cobalt (Co(2+)) were prepared by the ammoniacal precipitation method and the extent of doping was measured by ICP-OES. The
Cong Mao et al.
Biomedical materials (Bristol, England), 10(2), 025005-025005 (2015-03-26)
The ability of biomaterials to induce rapid vascular formation is critical in tissue regeneration. Combining recombinant angiogenic growth factors with bioengineered constructs have proven to be difficult due to several issues, including the instability of recombinant proteins, the need for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service