Skip to Content
Merck
All Photos(1)

Key Documents

09-1540

Sigma-Aldrich

Ethylene glycol

JIS special grade, ≥99.5%

Synonym(s):

1,2-Ethanediol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH2OH
CAS Number:
Molecular Weight:
62.07
Beilstein:
505945
MDL number:
UNSPSC Code:
12352137
PubChem Substance ID:

grade

JIS special grade

vapor density

2.1 (vs air)

vapor pressure

0.08 mmHg ( 20 °C)

Assay

≥99.5%

form

liquid

autoignition temp.

752 °F

expl. lim.

15.3 %

availability

available only in Japan

refractive index

n20/D 1.431 (lit.)

bp

195-198 °C

mp

−13 °C (lit.)

density

1.113 g/mL at 25 °C (lit.)

SMILES string

OCCO

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT RE 2 Oral

Target Organs

Kidney

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

231.8 °F

Flash Point(C)

111 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi Wang et al.
Journal of the American Chemical Society, 135(5), 1941-1951 (2013-01-16)
This article describes a robust method for the facile synthesis of small Ag nanocubes with edge lengths controlled in the range of 18-32 nm. The success of this new method relies on the substitution of ethylene glycol (EG)--the solvent most
Revital Goldshmid et al.
Methods (San Diego, Calif.), 84, 35-43 (2015-05-02)
A method for the preparation of suspension culture microcapsules used in the bioprocessing of human mesenchymal stem cells (hMSCs) is reported. The microcapsules are prepared from a semi-synthetic hydrogel comprising Pluronic®F127 conjugated to denatured fibrinogen. The Pluronic-fibrinogen adducts display a
Laetitia Galea et al.
Biomaterials, 34(27), 6388-6401 (2013-06-13)
Calcium phosphates (CaPs) are widely used as bone graft substitutes but are inherently brittle, hence restricting their use to mechanically protected environments. Combining them with a tough polymer matrix could potentially lead to a composite with load-bearing properties. However, the
Marco Scavini et al.
IUCrJ, 2(Pt 5), 511-522 (2015-08-26)
A new hierarchical approach is presented for elucidating the structural disorder in Ce1-x Gd x O2-x/2 solid solutions on different scale lengths. The primary goal of this investigation is to shed light on the relations between the short-range and the
Birgit Oberleitner et al.
Chemical communications (Cambridge, England), 49(16), 1615-1617 (2013-01-23)
This work describes an integrated approach for designing on demand Self-Assembled Monolayers (SAMs) on silicon oxides and particularly glass substrates for cell biology applications. Starting from commercially available compounds, the strategy relies on thiol-ene reaction and provides high quality SAMs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service