Skip to Content
Merck
All Photos(1)

Documents

01-4940

Sigma-Aldrich

Ammonium phosphate dibasic

JIS special grade, ≥99.0%

Synonym(s):

Ammonium hydrogenphosphate, Diammonium hydrogenphosphate, di-Ammonium hydrogenphosphate (sec)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)2HPO4
CAS Number:
Molecular Weight:
132.06
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

JIS special grade

Assay

≥99.0%

form

crystalline

availability

available only in Japan

pH

7.5-9.0 (25 °C, 132.1 g/L)

mp

155 °C (dec.) (lit.)

SMILES string

N.N.OP(O)(O)=O

InChI

1S/2H3N.H3O4P/c;;1-5(2,3)4/h2*1H3;(H3,1,2,3,4)

InChI key

MNNHAPBLZZVQHP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Catarina Barbosa et al.
International journal of food microbiology, 160(2), 87-93 (2012-11-28)
Sulphur-containing amino acids, cysteine and methionine, are generally found in very low concentrations in grape-juice. The objective of this study was to identify the effects of methionine on aroma compounds formation. Nitrogen source effects on growth, fermentative behaviour and aroma
Nathan K Deed et al.
Applied microbiology and biotechnology, 89(5), 1537-1549 (2011-01-20)
Two deletion mutants expected to be defective in nitrogen catabolite repression (NCR) were constructed in a commercial wine yeast background M2: a ure2 mutant and a dal80 gzf3 double mutant. Wild-type and both mutant strains were fermented in Sauvignon Blanc
Alicia Gutiérrez et al.
Food microbiology, 31(1), 25-32 (2012-04-06)
Nitrogen deficiencies in grape musts are one of the main causes of stuck or sluggish wine fermentations. Currently, the most common method for dealing with nitrogen-deficient fermentations is adding supplementary nitrogen (usually ammonium phosphate). However, it is important to know
Karima Achelhi et al.
Dalton transactions (Cambridge, England : 2003), 39(44), 10644-10651 (2010-10-05)
Organically-modified hydroxyapatite materials were synthesized through the addition of oxalic, succinic, adipic and citric acids to a calcium hydroxide solution before neutralization by ammonium dihydrogenphosphate. All carboxylic acids have a significant influence on apatite crystallinity and nanoparticle size, as indicated
Everest Akaraonye et al.
Biotechnology journal, 7(2), 293-303 (2011-12-08)
The main hindrance in the use of polyhydroxyalkanoates (PHAs) as a replacement for existing petroleum-based plastics is their high production cost. The carbon source accounts for 50% of the cost for PHA production. Thus, increasing the yield and productivity of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service