Merck
  • The kinetics of the swelling process and the release mechanisms of Coriandrum sativum L. essential oil from chitosan/alginate/inulin microcapsules.
The kinetics of the swelling process and the release mechanisms of Coriandrum sativum L. essential oil from chitosan/alginate/inulin microcapsules.
Food chemistry (2015-11-18)
Cristian Dima, Livia Pătraşcu, Alina Cantaragiu, Petru Alexe, Ştefan Dima
ABSTRACT
The encapsulation by spray drying method of coriander essential oil (CEO) in various materials (chitosan, alginate, chitosan/alginate, chitosan/inulin) was studied. The viscoelastic properties of the oil-in-water (O/W) emulsions and the characteristics of CEO-loaded microcapsules like morphology, moisture, wettability, solubility, flowability properties, swelling and release mechanisms were investigated. The chitosan microcapsules had a brain-like structure while the alginate and chitosan/alginate microcapsules are spherical with a smooth surface. The Compressibility Index (CI=29.09-32.25%) and Hausner Ratio (HR=1.38-1.44) values showed that all the microcapsules prepared correspond to the "poor" flowability powders group. The chitosan microcapsules exhibited the maximum release rate at pH 2.5 while the alginate microcapsules exhibited the maximum release rate at pH 6.5. Kinetics and mechanism of CEO release were studied using various mathematical models such as, zero order, first order, Higuchi model and Peppas model. The diffusional exponent (n) values of Peppas equation explains a non Fickian transport mechanism and diffusion or diffusion-swelling controlled process.
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Sigma-Aldrich
Alginic acid sodium salt from brown algae, low viscosity
Sigma-Aldrich
Chitosan, low molecular weight
Sigma-Aldrich
Chitosan, from shrimp shells, ≥75% (deacetylated)
Sigma-Aldrich
Alginic acid sodium salt from brown algae, Medium viscosity
Sigma-Aldrich
L-α-Phosphatidylcholine, from dried egg yolk, Type X-E, ≥40% (enzymatic)
Sigma-Aldrich
L-α-Phosphatidylcholine, from soybean, ≥99% (TLC), lyophilized powder
Sigma-Aldrich
L-α-Phosphatidylcholine, from soybean, Type IV-S, ≥30% (enzymatic)
Sigma-Aldrich
L-α-Phosphatidylcholine, egg yolk, Type XIII-E, 100 mg/mL in ethanol, solution
Sigma-Aldrich
L-α-Phosphatidylcholine, solution, from soybean, suitable for cell culture, Type III-S
Sigma-Aldrich
L-α-Phosphatidylcholine, egg yolk, Type XVI-E, ≥99% (TLC), lyophilized powder
Sigma-Aldrich
Inulin from dahlia tubers
Sigma-Aldrich
L-α-Phosphatidylcholine, egg yolk, Type XI-E, 100 mg/mL in chloroform, ≥99%, solution
Sigma-Aldrich
L-α-Phosphatidylcholine, hydrogenated, Type I-EH, ≥99%
Sigma-Aldrich
Chitosan, medium molecular weight
Sigma-Aldrich
Chitosan, from shrimp shells, practical grade
Sigma-Aldrich
Chitosan, high molecular weight
Sigma-Aldrich
Alginic acid sodium salt, powder
Sigma-Aldrich
Alginic acid sodium salt from brown algae, BioReagent, suitable for immobilization of micro-organisms
Sigma-Aldrich
L-α-Phosphatidylcholine, from soybean, Type II-S, 14-29% choline basis
Sigma-Aldrich
Citric acid monohydrate, JIS special grade, ≥99.5%
Sigma-Aldrich
Citric acid monohydrate, ≥99.5%, suitable for amino acid analysis
Sigma-Aldrich
Citric acid monohydrate, SAJ first grade, ≥99.5%
Sigma-Aldrich
Sodium alginate
Sigma-Aldrich
Chitosan from shrimp shells, low-viscous
Sigma-Aldrich
Sodium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
Citric acid monohydrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., buffer substance, 99.5-102%
Sigma-Aldrich
Sodium phosphate dibasic, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Inulin from chicory
Sigma-Aldrich
Citric acid monohydrate, ACS reagent, ≥99.0%