Skip to Content
Merck
All Photos(1)

Documents

W201502

Sigma-Aldrich

Sodium alginate

Synonym(s):

Alginic acid sodium salt from brown algae, Algin, Sodium alginate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2015
MDL number:
UNSPSC Code:
12164502
E Number:
E 401
NACRES:
NA.21

biological source

synthetic

grade

Halal

reg. compliance

FDA 21 CFR 117
FDA 21 CFR 150.161
FDA 21 CFR 173.310

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

odorless

InChI

1S/C6H10O7.Na/c7-1-2(8)4(5(10)11)13-6(12)3(1)9;/h1-4,6-9,12H,(H,10,11);/q;+1/p-1/t1-,2-,3-,4?,6+;/m0./s1

InChI key

MSXHSNHNTORCAW-MPGIDXPLSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Sodium alginate is the sodium salt of alginic acid, a polysaccharide found in brown algae. It is generally used as a stabilizer and thickener in the food industry.

Application

Sodium alginate can be used to prepare:
  • Cationized casein-based polyelectrolyte complex for fragrance-controlled release applications.
  • Aroma encapsulation or microcapsules for cosmetic applications.

Other Notes

A straight-chain, hydrophilic, colloidal, polyuronic acid composed of guluronic and mannuronic acid residues.

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Polyelectrolyte complex from cationized casein and sodium alginate for fragrance controlled release
Zhang Y, et al.
Colloids and Surfaces. B, Biointerfaces, 178, 439-444 (2019)
Preparation of aroma microcapsules with sodium alginate and tetradecylallyldimethylammonium bromide (TADAB) and its potential applications in cosmetics
Song J and Chen H
Flavour and Fragrance Journal, 33(2), 160-165 (2018)
A major constituent of brown algae for use in high-capacity Li-ion batteries.
Kovalenko I, et al.
Science, 334(6052), 75-79 (2011)
Encyclopedia of Food and Color Additives, 1, 67-70 (1997)
Núria Blanco-Cabra et al.
Scientific reports, 10(1), 9390-9390 (2020-06-12)
Aggregates of Pseudomonas aeruginosa form a protective barrier against antibiotics and the immune system. These barriers, known as biofilms, are associated with several infectious diseases. One of the main components of these biofilms is alginate, a homo- and hetero-polysaccharide that

Related Content

Tissue engineering fabricates tissues cultures from scaffolds, living cells, and biologically active molecules by simulating the microenvironment of the body to repair or replace damaged tissue.

Tissue engineering fabricates tissues cultures from scaffolds, living cells, and biologically active molecules by simulating the microenvironment of the body to repair or replace damaged tissue.

Tissue engineering fabricates tissues cultures from scaffolds, living cells, and biologically active molecules by simulating the microenvironment of the body to repair or replace damaged tissue.

Tissue engineering fabricates tissues cultures from scaffolds, living cells, and biologically active molecules by simulating the microenvironment of the body to repair or replace damaged tissue.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service