Skip to Content
Merck
All Photos(3)

Key Documents

I3754

Sigma-Aldrich

Inulin from dahlia tubers

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H10O5)n
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

dahlia tuber

form

powder

mol wt

Mr ~5000

color

white

mp

158 to 165 °C

solubility

water: 100 mg/mL, clear to hazy, colorless

SMILES string

OC[C@H]1O[C@](O)(CO)[C@@H](O)[C@@H]1O.OC[C@H]2OC(O[C@]3(CO)O[C@H](CO)[C@@H](O)[C@@H]3O)[C@H](O)[C@@H](O)[C@@H]2O

InChI

1S/C12H22O11.C6H12O6/c13-1-4-6(16)8(18)9(19)11(21-4)23-12(3-15)10(20)7(17)5(2-14)22-12;7-1-3-4(9)5(10)6(11,2-8)12-3/h4-11,13-20H,1-3H2;3-5,7-11H,1-2H2/t4-,5-,6-,7-,8+,9-,10+,11?,12+;3-,4-,5+,6-/m11/s1

InChI key

TUULCWHRUVPVEB-UGWHTDGHSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nathan Crook et al.
Cell host & microbe, 25(4), 499-512 (2019-03-31)
Probiotics are living microorganisms that are increasingly used as gastrointestinal therapeutics by virtue of their innate or engineered genetic function. Unlike abiotic therapeutics, probiotics can replicate in their intended site, subjecting their genomes and therapeutic properties to natural selection. We exposed
Grethel Teresa Choque Delgado et al.
Plant foods for human nutrition (Dordrecht, Netherlands), 68(3), 222-228 (2013-05-28)
Yacon (Smallanthus sonchifolius) is an Andean tuberous root that is regarded as a functional food given that it contains fructooligosaccharides (FOS), inulin and phenolic compounds. The consumption of FOS and inulin improves the growth of bifidobacteria in the colon, enhances
Alison N Beloshapka et al.
FEMS microbiology ecology, 84(3), 532-541 (2013-01-31)
Our objective was to determine the effects of feeding raw meat-based diets with or without inulin or yeast cell wall extract (YCW) on fecal microbial communities of dogs using 454 pyrosequencing. Six healthy female adult beagles (5.5 ± 0.5 years; 8.5 ± 0.5 kg) were randomly
Shiu-Ming Kuo
Advances in nutrition (Bethesda, Md.), 4(1), 16-28 (2013-01-16)
Fiber intake is critical for optimal health. This review covers the anti-inflammatory roles of fibers using results from human epidemiological observations, clinical trials, and animal studies. Fiber has body weight-related anti-inflammatory activity. With its lower energy density, a diet high
Jeroen van Arkel et al.
Carbohydrate polymers, 93(1), 48-56 (2013-03-08)
Fructan, a fructose polymer, is produced by many bacteria and plants. Fructan is used as carbohydrate reserve, and in bacteria also as protective outside layer. Chicory is a commercial fructan producing crop. The disadvantage of this crop is its fructan

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service