Skip to Content
Merck
All Photos(1)

Documents

71636

Sigma-Aldrich

Sodium phosphate dibasic

BioUltra, for molecular biology, ≥99.5% (T)

Synonym(s):

sec-Sodium phosphate, Disodium hydrogen phosphate, Disodium phosphate, Sodium hydrogenphosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2HPO4
CAS Number:
Molecular Weight:
141.96
EC Number:
MDL number:
UNSPSC Code:
12161700
eCl@ss:
38070211
PubChem Substance ID:
NACRES:
NA.25

grade

for molecular biology

product line

BioUltra

Assay

≥99.5% (T)

form

coarse powder
fine crystals
powder

impurities

DNases, none detected
RNases, none detected
insoluble matter, passes filter test
phosphatases, none detected
proteases, none detected
≤0.001% total nitrogen (N)

loss

≤0.1% loss on drying, 110 °C

pH

8.8-9.4 (25 °C, 0.5 M in H2O)

pKa (25 °C)

(1) 2.15, (2) 6.82, (3) 12.38 (phosphoric acid)

solubility

H2O: 0.5 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤10 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.2 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤10 mg/kg
Zn: ≤5 mg/kg

λ

0.5 M in H2O

UV absorption

λ: 260 nm Amax: ≤0.025
λ: 280 nm Amax: ≤0.020

SMILES string

[Na+].[Na+].[H]OP([O-])([O-])=O

InChI

1S/2Na.H3O4P/c;;1-5(2,3)4/h;;(H3,1,2,3,4)/q2*+1;/p-2

InChI key

BNIILDVGGAEEIG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Sodium phosphate dibasic has been used:
  • as a component in phosphate buffer
  • in the preparation of MgCl2 hybridization buffer
  • as a component in SSPE solution for the preparation of small RNA hybridisation buffer

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Design of biomimetic collagen matrices by reagent-free electron beam induced crosslinking: Structure-property relationships and cellular response
Riedel S, et al.
Materials & Design, 107606-107606 (2019)
Foundations of identifying individual chromosomes by imaging flow cytometry with applications in radiation biodosimetry
Beaton-Green LA, et al.
Methods, 112, 18-24 (2017)
Eva Várallyay et al.
Nature protocols, 3(2), 190-196 (2008-02-16)
MicroRNAs (miRNAs) are short, about 21 nucleotides in length, noncoding, regulatory RNA molecules representing a new layer in post-transcriptional regulation of gene expression. Intensive miRNA research has necessitated the development of effective miRNA detection methods such as northern analyses, quantitative
Kaisorn L Chaichana et al.
Journal of neuroscience methods, 180(1), 116-125 (2009-05-12)
The discoveries of neural (NSCs) and brain tumor stem cells (BTSCs) in the adult human brain and in brain tumors, respectively, have led to a new era in neuroscience research. These cells represent novel approaches to studying normal phenomena such
Janelle M Spinazzola et al.
Human molecular genetics, 24(9), 2470-2481 (2015-01-22)
Loss of gamma-sarcoglycan (γ-SG) induces muscle degeneration and signaling defects in response to mechanical load, and its absence is common to both Duchenne and limb girdle muscular dystrophies. Growing evidence suggests that aberrant signaling contributes to the disease pathology; however

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service