Skip to Content
Merck
All Photos(3)

Key Documents

P5638

Sigma-Aldrich

L-α-Phosphatidylcholine

from soybean, Type II-S, 14-29% choline basis

Synonym(s):

1,2-Diacyl-sn-glycero-3-phosphocholine, 3-sn-Phosphatidylcholine, L-α-Lecithin, Azolectin, PC

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Beilstein:
5209585
EC Number:
UNSPSC Code:
12352211
NACRES:
NA.25

biological source

soybean

Quality Level

type

Type II-S

form

powder or solid

concentration

14-29% (choline)

functional group

phospholipid

lipid type

phosphoglycerides

shipped in

ambient

storage temp.

2-8°C

SMILES string

[P](=O)([O-])(OC[C@H](OC(=O)CCCCCCC\C=C/C\C=C/CCCCC)COC(=O)CCCCCCCCCCCCCCC)OCC[N+](C)(C)C

InChI

1S/C42H80NO8P/c1-6-8-10-12-14-16-18-20-21-23-25-27-29-31-33-35-42(45)51-40(39-50-52(46,47)49-37-36-43(3,4)5)38-48-41(44)34-32-30-28-26-24-22-19-17-15-13-11-9-7-2/h14,16,20-21,40H,6-13,15,17-19,22-39H2,1-5H3/b16-14-,21-20-/t40-/m1/s1

InChI key

JLPULHDHAOZNQI-ZTIMHPMXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

P5638 is a crude extract of soybean phospholipids.

Typical lots of purified soybean phosphatidylcholine have fatty acid contents of approximately 13% C16:0 (palmitic), 4% C18:0 (stearic), 10% C18:1(oleic), 64% C18:2 (linoleic), and 6% 18:3 (linolenic) with other fatty acids being minor contributors. This would calculate to an average molecular weight of approximately 776.

Purified phosphatidylcholine is soluble in chloroform and ethanol.

Other major phospholipid components include phosphatidylethanolamine and inositol phosphatides. Other phospholipids and neutral lipids such as triglycerides, tocopherols, and sterols may be present. Carbohydrates may also be present.

A procedure for determination of the amount of oxidation of egg phosphatidylcholine in a liposome preparation by measurement of the oxidation index has been published. The oxidation index is the ratio of the absorbance at 233 nm to the absorbance at 215 nm. The latter wavelength was chosen since there is little contribution of the fatty acid carbonyl to the absorbance at this wavelength, allowing Beer′s Law to be followed.

A method of determining choline content has been published.

Biochem/physiol Actions

A major structural phospholipid in brain, comprising approx. 15% of total lipid; primarily localized to gray matter.
Phosphatidylcholine is the major membrane phospholipid in eukaryotic cells. In addition to being the major structural component of cellular membranes, phosphatidylcholine serves as a reservoir for several lipid messengers. It is the source of the bioactive lipids lysophosphatidylcholine, phosphatidic acid, diacylglycerol, lysophosphatidylcholine, platelet activating factor, and arachidonic acid.

Phosphatidylcholine is used for preparation of vesicle suspensions, commonly called liposomes, or as monolayers. Monolayers have been formed using a solution of 1% soybean phosphatidylcholine in hexane.

Valinomycin-induced changes in membrane potentials of red blood cell and phospholipid (phosphatidylcholine from egg yolk plus cholesterol) vesicle suspensions have been measured using positively-charged, cyanine dyes that fluorimetrically responded to the change in potential.

Purified rhodopsin has been incorporated into soybean phosphatidylcholine vesicles.

Quality

P5638 is a crude soy extract that contains many other lipids.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

P M Guy et al.
Biochemistry, 29(30), 6954-6964 (1990-07-31)
The intrinsic tryptophan fluorescence of the alpha subunit of transducin (alpha T) has been shown to be sensitive to the binding of guanine nucleotides, with the fluorescence being enhanced by as much as 2-fold upon the binding of GTP or
A simplified and specific method for the estimation of choline.
C J ACKERMAN et al.
Analytical biochemistry, 1, 327-336 (1960-12-10)
Austin J Rice et al.
The Journal of biological chemistry, 288(29), 21228-21235 (2013-05-28)
In bacteria, ATP-binding cassette (ABC) transporters are vital for the uptake of nutrients and cofactors. Based on differences in structure and activity, ABC importers are divided into two types. Type I transporters have been well studied and employ a tightly
Liposomes in Gene Delivery
Lasic, D.D. et al.
CRC Press (1997)
Burcu Ustuner et al.
Animal reproduction science, 164, 97-104 (2015-12-20)
The aim of the current study was to evaluate the effects of different concentrations of rainbow trout seminal plasma (RTSP) (0.1%, 1% and 10%) in extenders containing either egg yolk or lecithin for use in Awassi ram semen cryopreservation. Pooled

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service