Merck
Zwitterionic red fluorescent polymeric nanoparticles for cell imaging.
Macromolecular bioscience (2014-06-14)
Haiyin Li, Xiqi Zhang, Xiaoyong Zhang, Bin Yang, Yang Yang, Zengfang Huang, Yen Wei
ABSTRACT
Phospholipid monomer and aggregation-induced emission (AIE) dye-based dimers are incorporated via reversible addition-fragmentation transfer polymerization to afford cross-linked zwitterionic fluorescent copolymers. Such copolymers are prone to self-assembly into fluorescent polymeric nanoparticles (FPNs) in physiological solution due to their amphiphilic nature. Characterization of these red FPNs by Fourier-transform infrared spectroscopy and X-ray photoelectron spectroscopy indicates the successful preparation of these zwitterionic copolymers. UV-visible absorption spectroscopy, fluorescence spectroscopy, transmission electron microscopy, and dynamic light scattering are performed to demonstrate the bright red fluorescence of the FPNs and their stable dispersibilities, even below the critical micelle concentration in physiological solution. Finally, studies of the biocompatibility and cell uptake behavior of the FPNs are conducted and show excellent biocompatibility for cell imaging application.
Sigma-Aldrich
Tetrakis(triphenylphosphine)palladium(0), 99%
Sigma-Aldrich
4-Vinylphenylboronic acid, ≥95%
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
2-Methacryloyloxyethyl phosphorylcholine, contains ≤100 ppm MEHQ as inhibitor, 97%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Tetrakis(triphenylphosphine)palladium(0), ≥99.99% trace metals basis
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, 1.0 M in methanol
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, technical, ~40% in H2O (~1.5 M)
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, 40 wt. % in H2O
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, 53.5-56.5% in H2O
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Supelco
Tetrabutylammonium hydroxide solution, ~40% in water, suitable for ion chromatography
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%