Skip to Content
Merck
All Photos(3)

Key Documents

230189

Sigma-Aldrich

Tetrabutylammonium hydroxide solution

greener alternative

1.0 M in methanol

Synonym(s):

N,N,N-Tributyl-1-butanaminium hydroxide, TBAH 40, Tetra-n-butylammonium hydroxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)4N(OH)
CAS Number:
Molecular Weight:
259.47
Beilstein:
3571228
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

form

liquid

Quality Level

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

1.0 M in methanol

refractive index

n20/D 1.3775

density

0.83 g/mL at 25 °C

functional group

amine

greener alternative category

SMILES string

[OH-].CCCC[N+](CCCC)(CCCC)CCCC

InChI

1S/C16H36N.H2O/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;/h5-16H2,1-4H3;1H2/q+1;/p-1

InChI key

VDZOOKBUILJEDG-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Tetrabutylammonium hydroxide is a quaternary ammonium salt mainly used as a strong base and phase-transfer catalyst.
Tetrabutylammonium hydroxide solution serves as a phase-transfer catalyst in the transesterification of soybean oil.


We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product is highly effcient and enhance catalytic efficiency. Click here for more information.

Application

Tetrabutylammonium hydroxide solution (1M in methanol) may be used as a base to synthesize ring methyl and methylene deuteriated porphyrins and metalloporphyrins. It may also be used as an initiator of graft polymerization between pivalolactone and poly(ethylene-co-vinyl acetate-co-methacrylic acid).

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rapid transesterification of soybean oil with phase transfer catalysts
Zhang Y, et al.
Applied Catalysis A: General, 366(1), 176-183 (2009)
Block and Graft Copolymers of Pivalolactone. 3. Grafts on Ethylene Copolymers
Sundet SA
Macromolecules, 11(1), 146-149 (1978)
Tetrabutylammonium Hydroxide
Bos ME
e-EROS Encyclopedia of Reagents for Organic Synthesis. (2001)
Rapid base-catalyzed deuterium exchange at the ring-adjacent methyl and methylene positions of octaalkyl and natural-derivative porphyrins and metalloporphyrins.
Godziela GM, et al.
Inorganic Chemistry, 25(23), 4286-4288 (1986)
Tetrabutylammonium hydroxide as titrant in nonaqueous media.
Harlow GA, et al.
Analytical Chemistry, 28(5), 787-791 (1956)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service