Skip to Content
Merck
All Photos(2)

Key Documents

697265

Sigma-Aldrich

Tetrakis(triphenylphosphine)palladium(0)

≥99.99% trace metals basis

Synonym(s):

Palladium-tetrakis(triphenylphosphine), Pd(PPh3)4

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pd[(C6H5)3P]4
CAS Number:
Molecular Weight:
1155.56
Beilstein:
6704828
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.99% trace metals basis

form

crystals

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

storage temp.

2-8°C

SMILES string

[Pd].c1ccc(cc1)P(c2ccccc2)c3ccccc3.c4ccc(cc4)P(c5ccccc5)c6ccccc6.c7ccc(cc7)P(c8ccccc8)c9ccccc9.c%10ccc(cc%10)P(c%11ccccc%11)c%12ccccc%12

InChI

1S/4C18H15P.Pd/c4*1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;/h4*1-15H;

InChI key

NFHFRUOZVGFOOS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenhan Zhang et al.
Angewandte Chemie (International ed. in English), 53(34), 8980-8984 (2014-07-01)
tert-Butoxyacetylene is shown to undergo Sonogashira coupling with aryl iodides to yield aryl-substituted tert-butyl ynol ethers. These intermediates participate in a [1,5]-hydride shift, which results in the extrusion of isobutylene and the generation of aryl ketenes. The ketenes are trapped
Chanchal Chakraborty et al.
ACS applied materials & interfaces, 7(34), 19034-19042 (2015-08-19)
A platinum(II)-based, luminescent, metallo-supramolecular polymer (PolyPtL1) having an inherent dipole moment was synthesized via complexation of Pt(II) ions with an asymmetric ligand L1, containing terpyridyl and pyridyl moieties. The synthesized ligand and polymer were well characterized by various NMR techniques
Debabrata Jana et al.
Organic & biomolecular chemistry, 13(43), 10663-10674 (2015-09-09)
Four pyrene-vinyl-tetraphenylethylene based conjugated materials were synthesized and characterized by FT-IR, NMR, and mass spectroscopy. The photophysical (including absorption, fluorescence, and fluorescence lifetime) and aggregation properties in tetrahydrofuran were investigated. The photophysical and aggregation behavior depends on the spacer, substituent
Shouchun Yin et al.
Soft matter, 11(22), 4424-4429 (2015-05-01)
In this paper, a pyrene moiety is incorporated into a bolaamphiphile to form a novel molecule denoted PRB. Above the critical micelle concentration, PRB forms nanodisks in the aqueous solution. The addition of acetate ions induces a morphological change in
Haiyin Li et al.
Macromolecular bioscience, 14(10), 1361-1367 (2014-06-14)
Phospholipid monomer and aggregation-induced emission (AIE) dye-based dimers are incorporated via reversible addition-fragmentation transfer polymerization to afford cross-linked zwitterionic fluorescent copolymers. Such copolymers are prone to self-assembly into fluorescent polymeric nanoparticles (FPNs) in physiological solution due to their amphiphilic nature.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service