Skip to Content
Merck
All Photos(1)

Key Documents

82762

Supelco

Methanol

analytical standard

Synonym(s):

Methyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OH
CAS Number:
Molecular Weight:
32.04
Beilstein:
1098229
EC Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

1.11 (vs air)

vapor pressure

410 mmHg ( 50 °C)
97.68 mmHg ( 20 °C)

Assay

≥99.9% (GC)

form

liquid

autoignition temp.

725 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

36 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.329 (lit.)
n20/D 1.329

bp

64.7 °C (lit.)

mp

−98 °C (lit.)

density

0.791 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

SMILES string

CO

InChI

1S/CH4O/c1-2/h2H,1H3

InChI key

OKKJLVBELUTLKV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methanol is a primary alcohol, which is commercially manufactured from methane. It can also be obtained via pyrolysis of wood. It can find applications in organic synthesis reactions, as a highly polar solvent.
This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Application

Methanol may be used as an analytical standard for the determination of the analyte content in biodiesel, wine, and biological fluids by gas chromatography (GC) technique. It can also be used as a solvent/eluent for the preparation of various standard solutions and the extraction of analytes for their subsequent determination using analytical techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The calculation of retention and selectivity in reversed-phase liquid chromatography II. Methanol-water eluents
Galushko SV
Chromatographia, 36(1), 39-42 (1993)
Methanol Fuel Cell Systems: Advancing Towards Commercialization (2016)
Determination of di-and tributyltin in sediment and microbial biofilms using acidified methanol extraction, sodium borohydride derivatization and gas chromatography with flame photometric detection
Matthias CL, et al.
International Journal of Environmental Analytical Chemistry, 35(2), 61-68 (1989)
Chromatography separation, 1H NMR analysis and bioautography screening of methanol extract of Excoecaria agallocha L. from Bhitarkanika, Orissa, India
Patra JK, et al.
Asian Pacific Journal of Tropical Biomedicine, 2(1), S50-S56 (2012)
Gas chromatographic analysis of methyl formate and application in methanol poisoning cases
Fraser AD and MacNeil W
Journal of Analytical Toxicology, 13(2), 73-76 (1989)

Articles

Learn about analyzing organic volatile impurities (OVIs) in pharmaceuticals using SH-GC, focusing on suitable solvents and ensuring compliance.

Learn about analyzing organic volatile impurities (OVIs) in pharmaceuticals using SH-GC, focusing on suitable solvents and ensuring compliance.

Learn about analyzing organic volatile impurities (OVIs) in pharmaceuticals using SH-GC, focusing on suitable solvents and ensuring compliance.

Learn about analyzing organic volatile impurities (OVIs) in pharmaceuticals using SH-GC, focusing on suitable solvents and ensuring compliance.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service