Skip to Content
Merck
All Photos(1)

Key Documents

632546

Supelco

Methanol solution

≥99.5%, suitable for UHPLC, mass spectrometry (MS), HPLC, contains 0.10% (v/v) formic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH4O
CAS Number:
Molecular Weight:
32.04
Beilstein:
3662861
EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:
grade:
HPLC grade
technique(s):
HPLC: suitable
LC/MS: suitable
UHPLC: suitable
mass spectrometry (MS): suitable
bp:
65.4 °C
vapor pressure:
410 mmHg

Product Name

Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%

grade

HPLC grade

Quality Level

vapor density

1.11 (vs air)

vapor pressure

410 mmHg

Assay

≥99.5%

form

liquid

contains

0.10 % (v/v) formic acid

expl. lim.

5.5-44 %

technique(s)

HPLC: suitable
LC/MS: suitable
UHPLC: suitable
mass spectrometry (MS): suitable

transmittance

230 nm, ≥10%
254 nm, ≥90%

bp

65.4 °C

mp

-98 °C

cation traces

Ca: ≤0.5 ppm
K: ≤0.5 ppm
Mg: ≤0.5 ppm
Na: ≤2 ppm

HPLC-gradient

≤10 mAU at 254 nm
≤20 mAU at 210 nm

suitability

suitable for LC-MS

format

mixture

SMILES string

CO

InChI

1S/CH4O/c1-2/h2H,1H3

InChI key

OKKJLVBELUTLKV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This pre-blended solvent eliminates time-consuming mobile phase preparation as well as lost sample information and instrument downtime caused by impure mobile phases. For consistent baselines and minimal background noise, these blends are prepared with ultra-pure acids such as formic acid.

Application

Designed for use with HPLC and LC-MS.

Packaging

M-Bottle for Solvents
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!

Preparation Note

Product filtered through a 0.2 μm filter

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adduct formation in quantitative bioanalysis: effect of ionization conditions on paclitaxel.
Mortier KA, et al.
Journal of the American Society For Mass Spectrometry, 15(4), 585-592 (2004)
Kentaro Fujiwara et al.
EJNMMI research, 4, 29-29 (2014-07-10)
ROBO1 is a membrane protein that functions in axon guidance. ROBO1 contributes to tumour metastasis and angiogenesis and may have potential as a target protein of immunotherapy because ROBO1 is specifically expressed at high levels in hepatocellular carcinoma. In this
Haruka Oda et al.
Biomaterials, 56, 86-91 (2015-05-03)
A large number of lineage-committed progenitor cells are required for advanced regenerative medicine based on cell engineering. Due to their ability to differentiate into multiple cells lines, multipotent stem cells have emerged as a vital source for generating transplantable cells
Timo Heidt et al.
Circulation research, 115(2), 284-295 (2014-05-03)
Macrophages populate the steady-state myocardium. Previously, all macrophages were thought to arise from monocytes; however, it emerged that, in several organs, tissue-resident macrophages may self-maintain through local proliferation. Our aim was to study the contribution of monocytes to cardiac-resident macrophages
Panagiotis Ntziachristos et al.
Nature, 514(7523), 513-517 (2014-08-19)
T-cell acute lymphoblastic leukaemia (T-ALL) is a haematological malignancy with a dismal overall prognosis, including a relapse rate of up to 25%, mainly because of the lack of non-cytotoxic targeted therapy options. Drugs that target the function of key epigenetic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service