Merck
  • Erythropoietin responsive cardiomyogenic cells contribute to heart repair post myocardial infarction.
Erythropoietin responsive cardiomyogenic cells contribute to heart repair post myocardial infarction.
Stem cells (Dayton, Ohio) (2014-05-09)
Maria Patapia Zafiriou, Claudia Noack, Bernhard Unsöld, Michael Didie, Elena Pavlova, Henrike J Fischer, Holger M Reichardt, Martin W Bergmann, Ali El-Armouche, Wolfram-Hubertus Zimmermann, Laura Cecilia Zelarayan
ABSTRACT
The role of erythropoietin (Epo) in myocardial repair after infarction remains inconclusive. We observed high Epo receptor (EPOR) expression in cardiac progenitor cells (CPCs). Therefore, we aimed to characterize these cells and elucidate their contribution to myocardial regeneration on Epo stimulation. High EPOR expression was detected during murine embryonic heart development followed by a marked decrease until adulthood. EPOR-positive cells in the adult heart were identified in a CPC-enriched cell population and showed coexpression of stem, mesenchymal, endothelial, and cardiomyogenic cell markers. We focused on the population coexpressing early (TBX5, NKX2.5) and definitive (myosin heavy chain [MHC], cardiac Troponin T [cTNT]) cardiomyocyte markers. Epo increased their proliferation and thus were designated as Epo-responsive MHC expressing cells (EMCs). In vitro, EMCs proliferated and partially differentiated toward cardiomyocyte-like cells. Repetitive Epo administration in mice with myocardial infarction (cumulative dose 4 IU/g) resulted in an increase in cardiac EMCs and cTNT-positive cells in the infarcted area. This was further accompanied by a significant preservation of cardiac function when compared with control mice. Our study characterized an EPO-responsive MHC-expressing cell population in the adult heart. Repetitive, moderate-dose Epo treatment enhanced the proliferation of EMCs resulting in preservation of post-ischemic cardiac function.
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
L-Ascorbic acid, 99%
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
Sodium pyruvate, SAJ special grade, ≥95.0%
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
L-Glutamine