Skip to Content
Merck
All Photos(1)

Documents

A4403

Sigma-Aldrich

L-Ascorbic acid

powder, suitable for cell culture, γ-irradiated

Synonym(s):

L-Threoascorbic acid, Antiscorbutic factor, Vitamin C

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8O6
CAS Number:
Molecular Weight:
176.12
Beilstein:
84272
EC Number:
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.75

biological source

synthetic (organic)

Quality Level

sterility

γ-irradiated

form

powder

mol wt

Mw 176.12 g/mol

technique(s)

cell culture | insect: suitable
cell culture | mammalian: suitable
cell culture | plant: suitable

color

white to off-white

pH

1.0-2.5 (25 °C, 176 g/L in water)

mp

190-194 °C (dec.)

solubility

H2O: 10 mg/mL

SMILES string

OC([C@]([C@@H](O)CO)([H])O1)=C(O)C1=O

InChI

1S/C6H8O6/c7-1-2(8)5-3(9)4(10)6(11)12-5/h2,5,7-10H,1H2/t2-,5+/m0/s1

InChI key

CIWBSHSKHKDKBQ-JLAZNSOCSA-N

Gene Information

human ... SLC23A2(9962)

Looking for similar products? Visit Product Comparison Guide

General description

L-ascorbic acid is an unsaturated γ-lactone ring containing an enediol configuration (–COH=COH) attached with a carbonyl group.

Application

For use in cell culture applications as an antioxidant and to regenerate tocopherol.
L-ascorbic acid has been used:
  • in an osteogenic supplement for the growth of cells
  • as a component in the differentiation media
  • in WS-MSCs (Werner syndrome-mesenchymal stem cells) to study rescuing properties of ascorbic acid on aging defects

Biochem/physiol Actions

L-ascorbic acid mainly exhibits antioxidant properties. It protects plants from oxidative stress and mammals from diseases associated with oxidative stress. L-ascorbic acid mainly protects from hydroxyl radicals, superoxide and singlet oxygen. In addition, it also reduces the membrane-linked antioxidant α-tocopherol (oxidised form). L-Ascorbic acid enhances endothelium-dependent vasodilation in various disorders, including diabetes, coronary artery disease, hypertension and chronic heart failure.

recommended

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Heller et al.
The Journal of biological chemistry, 274(12), 8254-8260 (1999-03-13)
Ascorbic acid has been shown to enhance impaired endothelium-dependent vasodilation in patients with atherosclerosis by a mechanism that is thought to involve protection of nitric oxide (NO) from inactivation by free oxygen radicals. The present study in human endothelial cells
Plant L-ascorbic acid: chemistry, function, metabolism, bioavailability and effects of processing.
Davey MW
Journal of the Science of Food and Agriculture, 80, 825-860 (2000)
Xi Wang et al.
Archives of oral biology, 57(9), 1231-1240 (2012-03-30)
This study focused on the characterization of stem cells from human exfoliated deciduous teeth (SHED) in comparison with dental pulp stem cells (DPSCs) to certify SHED as a key element in tissue engineering. In the present study, SHED and DPSCs
P L Conklin et al.
Plant physiology, 115(3), 1277-1285 (1997-12-09)
The biosynthesis of L-ascorbic acid (vitamin C) is not well understood in plants. The ozone-sensitive Arabidopsis thaliana mutant vitamin c-1 (vtc1; formerly known as soz1) is deficient in ascorbic acid, accumulating approximately 30% of wild-type levels. This deficiency could result
Ying Li et al.
Protein & cell, 7(7), 478-488 (2016-06-09)
Werner syndrome (WS) is a premature aging disorder that mainly affects tissues derived from mesoderm. We have recently developed a novel human WS model using WRN-deficient human mesenchymal stem cells (MSCs). This model recapitulates many phenotypic features of WS. Based

Articles

Importance and uses of ascorbate in serum-free eukaryotic, including hybridoma and Chinese Hamster Ovary (CHO) cell, cultures

Importance and uses of ascorbate in serum-free eukaryotic, including hybridoma and Chinese Hamster Ovary (CHO) cell, cultures

Importance and uses of ascorbate in serum-free eukaryotic, including hybridoma and Chinese Hamster Ovary (CHO) cell, cultures

Importance and uses of ascorbate in serum-free eukaryotic, including hybridoma and Chinese Hamster Ovary (CHO) cell, cultures

See All

Protocols

TrueGel3D® Hydrogel Plate protocol guides high-throughput culture of human adipose MSCs for screening applications.

TrueGel3D® Hydrogel Plate protocol guides high-throughput culture of human adipose MSCs for screening applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service