Skip to Content
Merck
All Photos(4)

Key Documents

P5280

Sigma-Aldrich

Sodium pyruvate

Sodium pyruvate

powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%

Synonym(s):

α-Ketopropionic acid sodium salt, 2-Oxopropanoic acid sodium salt, Pyruvic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCOONa
CAS Number:
Molecular Weight:
110.04
Beilstein:
3568341
EC Number:
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.71
Pricing and availability is not currently available.

product line

BioReagent

Quality Level

Assay

≥99%

form

powder

technique(s)

cell culture | insect: suitable
cell culture | mammalian: suitable

mp

>300 °C (lit.)

solubility

H2O: 100 mg/mL

density

1.78 at 20 °C

application(s)

metabolomics

storage temp.

2-8°C

SMILES string

[Na+].CC(=O)C([O-])=O

InChI

1S/C3H4O3.Na/c1-2(4)3(5)6;/h1H3,(H,5,6);/q;+1/p-1

InChI key

DAEPDZWVDSPTHF-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium pyruvate, derived from pyruvic acid, finds extensive use in cell biology, cell culture, and biochemical research. As the end-product of the glycolysis pathway, where glucose is converted into pyruvate with the energetic production of ATP (Adenosine Triphosphate), pyruvate serves as an intermediary in numerous metabolic pathways, including sugar metabolism. While cells naturally synthesize sodium pyruvate as an intermediate metabolite in glycolysis, it isn′t universally necessary as a supplement for all cell cultures. However, if cells have been cultivated in a medium enriched with sodium pyruvate, it′s advisable to continue its use as cell growth may diminish without it.Sodium pyruvate is frequently incorporated into cell culture formulations to fortify broths and media, enhancing cell survival by providing an easily-accessible carbohydrate energy source alongside glucose. It′s often added to the growth medium, particularly for rapidly proliferating cells like tumor cells. Furthermore, it participates in amino acid metabolism and initiates the Krebs cycle. Cells can readily utilize sodium pyruvate as a carbon source for energy production and crucial metabolic pathways, eliminating the need for biosynthetic production from glucose or amino acids.Certain cell lines require supplemental pyruvate in the culture media due to their inability to convert glucose or amino acids into pyruvate. Additionally, sodium pyruvate serves as a free radical scavenger, effectively quenching reactive oxygen species (ROS) and contributing to cellular health and viability.

Application

Sodium pyruvate has been used:
  • to culture MCF7 ρ0 (human breast cancer cell line) cells as parent cells
  • as a supplement in extracellular flux (XF) assay media to incubate cells for measuring oxygen consumption rate (OCR)
  • as a supplement in minimum essential medium Eagle to maintain breast cancer (MCF-7) cells
  • as a supplement in Dulbecco′s modified Eagle medium (DMEM)-based medium and modified Eagle medium (MEM)-α for testis cell culture to evaluate the effect of medium composition on in vitro SSC maintenance

Biochem/physiol Actions

Sodium Pyruvate has shown antioxidant properties and protective effects against oxygen radicals. Pyruvate is produced as part of glycolysis and is an intermediate in many metabolic pathways. It can be converted into acetyl CoA and enter the TCA Cycle.
Sodium pyruvate can be used as a component in culture broth and media. It exhibits protective effects in various animal models of ischemia-reperfusion injury, hemorrhagic shock, and septic shock when given exogenously.

Features and Benefits

  • Cell culture and insect cell culture tested

  • Versatile and adaptable for a wide variety of laboratory and research applications
  • BioReagent Grade Sodium succinate for your Cell Biology and Biochemical research
  • Tested for Mammalian and Insect cell culture applications

Other Notes

For additional information on our range of Biochemicals, please complete this form.

comparable product

Product No.
Description
Pricing

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1B

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei Yang et al.
Nature communications, 5, 3818-3818 (2014-05-03)
Polycomb Repressive Complex 1 and histone H2A ubiquitination (ubH2A) contribute to embryonic stem cell (ESC) pluripotency by repressing lineage-specific gene expression. However, whether active deubiquitination co-regulates ubH2A levels in ESCs and during differentiation is not known. Here we report that
Rekin's Janky et al.
PLoS computational biology, 10(7), e1003731-e1003731 (2014-07-25)
Identifying master regulators of biological processes and mapping their downstream gene networks are key challenges in systems biology. We developed a computational method, called iRegulon, to reverse-engineer the transcriptional regulatory network underlying a co-expressed gene set using cis-regulatory sequence analysis.
Philipp Müller et al.
Science translational medicine, 7(315), 315ra188-315ra188 (2015-11-27)
Targeted drug delivery with antibody-drug conjugates such as the HER2-directed ado-trastuzumab emtansine (T-DM1) has emerged as a powerful strategy for cancer therapy. We show that T-DM1 is particularly effective in eliciting antitumor immunity in patients with early breast cancer (WSG-ADAPT
Athanasia E Christakou et al.
Lab on a chip, 15(15), 3222-3231 (2015-07-02)
We demonstrate a simple method for three-dimensional (3D) cell culture controlled by ultrasonic standing waves in a multi-well microplate. The method gently arranges cells in a suspension into a single aggregate in each well of the microplate and, by this
Philipp Müller et al.
Cancer immunology research, 2(8), 741-755 (2014-06-12)
Antibody-drug conjugates (ADC) are emerging as powerful treatment strategies with outstanding target-specificity and high therapeutic activity in patients with cancer. Brentuximab vedotin represents a first-in-class ADC directed against CD30(+) malignancies. We hypothesized that its sustained clinical responses could be related

Articles

Warburg effect enhances glucose to lactate conversion in tumor cells, regardless of oxygen levels; impacting cancer metabolism since 1924.

Warburg effect enhances glucose to lactate conversion in tumor cells, regardless of oxygen levels; impacting cancer metabolism since 1924.

Warburg effect enhances glucose to lactate conversion in tumor cells, regardless of oxygen levels; impacting cancer metabolism since 1924.

Warburg effect enhances glucose to lactate conversion in tumor cells, regardless of oxygen levels; impacting cancer metabolism since 1924.

See All

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service