Skip to Content
Merck
All Photos(5)

Key Documents

A7506

Sigma-Aldrich

L-Ascorbic acid

reagent grade, crystalline

Synonym(s):

L-Threoascorbic acid, Antiscorbutic factor, Vitamin C

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8O6
CAS Number:
Molecular Weight:
176.12
Beilstein:
84272
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26
Pricing and availability is not currently available.

biological source

synthetic

grade

reagent grade

Assay

≥98% ( with iodine, titration)

form

crystalline

technique(s)

HPLC: suitable
cell culture | stem cell: suitable

color

white to slightly yellow

pH

1.0-2.5 (25 °C, 176 g/L in water)

mp

190-194 °C (dec.)

solubility

water: soluble 50 mg/mL

storage temp.

room temp

SMILES string

OC([C@]([C@@H](O)CO)([H])O1)=C(O)C1=O

InChI

1S/C6H8O6/c7-1-2(8)5-3(9)4(10)6(11)12-5/h2,5,7-10H,1H2/t2-,5+/m0/s1

InChI key

CIWBSHSKHKDKBQ-JLAZNSOCSA-N

Gene Information

human ... SLC23A2(9962)

Looking for similar products? Visit Product Comparison Guide

General description

L-Ascorbic acid biosynthetic pathway varies in plants, animals and fungi. It is synthesized from L-gulono-1,4-lactone by the action of enzyme L-gulono-1,4-lactone oxidase (GLO).[1]

Application

L-Ascorbic acid has been used:
  • as a chondrogenic medium component for culturing bone-derived mesenchymal stem cells (BMSCs)(149)
  • as a reference standard in high-performance liquid chromatography system (HPLC)(150)
  • to test its effect on human gastric cancer GES-1 and AGS cells(151)

Biochem/physiol Actions

Ascorbic Acid, also known as Vitamin C, is a six-carbon lactone produced by plants and some animal species but not by humans and other primates. Ascorbic acid functions as an enzymatic cofactor for multiple enzymes, serving as an electron donor for monooxygenases and dioxygenases. Ascorbic acid also functions as a powerful antioxidant, particularly in regards to reactive oxygen species.
L-Ascorbic acid deficiency is associated with lack of L-gulono-1,4-lactone oxidase (GLO) enzyme.[1] It elicits antioxidant functionality and gets oxidized to dehydroascorbate via ascorbyl radical intermediate. L-Ascorbic acid is essential for maintaining the redox balance.[2] It is a cofactor for dopamine-β-hydroxylase and favors T cell proliferation, protects immune system and provides relief in common cold.[3]

Caution

May darken in storage.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vitamin C in health and disease: its role in the metabolism of cells and redox state in the brain
Figueroa-Mendez R and Rivas-Arancibia, S
Frontiers in Physiology, 6, 397-397 (2015)
L-Ascorbic acid in organic synthesis: an overview
Tripathi RP, et al.
Current Organic Chemistry, 13(1), 99-122 (2009)
Vitamin C in disease prevention and cure: an overview
Chambial S, et al.
Indian Journal of Clinical Biochemistry : IJCB, 28(4), 314-328 (2013)
Takashi Yoshida et al.
Nature chemical biology, 2(11), 596-607 (2006-09-26)
Transient receptor potential (TRP) proteins form plasma-membrane cation channels that act as sensors for diverse cellular stimuli. Here, we report a novel activation mechanism mediated by cysteine S-nitrosylation in TRP channels. Recombinant TRPC1, TRPC4, TRPC5, TRPV1, TRPV3 and TRPV4 of
Jennifer R Brum et al.
Science (New York, N.Y.), 348(6237), 1261498-1261498 (2015-05-23)
Viruses influence ecosystems by modulating microbial population size, diversity, metabolic outputs, and gene flow. Here, we use quantitative double-stranded DNA (dsDNA) viral-fraction metagenomes (viromes) and whole viral community morphological data sets from 43 Tara Oceans expedition samples to assess viral

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service