Skip to Content
Merck
All Photos(1)

Documents

46951

Sigma-Aldrich

Fluorescein isothiocyanate isomer I

≥97.5% (HPLC)

Synonym(s):

FITC, Fluorescein 5-isothiocyanate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H11NO5S
CAS Number:
Molecular Weight:
389.38
Beilstein:
1407295
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

Assay

≥97.5% (HPLC)

form

powder

impurities

≤1.5% Isomer II

mp

>360 °C (lit.)

solubility

acetone: soluble 0.1 g/10 mL

fluorescence

λex 492 nm; λem 518 nm (green)
λex 492 nm; λem 518 nm in 0.1 M phosphate pH 8.0

storage temp.

2-8°C

SMILES string

Oc1ccc2c(Oc3cc(O)ccc3C24OC(=O)c5cc(ccc45)N=C=S)c1

InChI

1S/C21H11NO5S/c23-12-2-5-16-18(8-12)26-19-9-13(24)3-6-17(19)21(16)15-4-1-11(22-10-28)7-14(15)20(25)27-21/h1-9,23-24H

InChI key

MHMNJMPURVTYEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reagent for the FITC labeling of proteins; Microsequencing of proteins and peptides (HPLC)

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eddie A James et al.
Arthritis & rheumatology (Hoboken, N.J.), 66(7), 1712-1722 (2014-03-26)
Rheumatoid arthritis (RA) is thought to be a T cell-mediated disease, based on its strong association with HLA class II alleles, clinical responsiveness to T cell-directed therapies, and the presence of CD4+ T cells in rheumatoid joints. The presence of
T Matthes et al.
Leukemia, 29(9), 1901-1908 (2015-03-11)
Multiple myeloma (MM) is a non-curable tumor developing in the bone marrow (BM). The BM microenvironment rich in hematopoietic precursors is suspected to have a role in MM development. Here we show that a proliferation-inducing ligand (APRIL) mediated in vivo
Aram Ghalali et al.
Carcinogenesis, 35(7), 1547-1555 (2014-01-24)
Epidemiological studies indicate that statins, cholesterol-lowering drugs, prevent aggressive prostate cancer and other types of cancer. Employing essentially non-prostate cell lines, we previously showed that statins rapidly downregulate nuclear levels of phosphorylated Akt via P2X7, a purinergic receptor recently implicated
Fangming Xiu et al.
Shock (Augusta, Ga.), 44(1), 44-51 (2015-05-07)
Relatively little is known about the effects of hepatocytes on hepatic macrophages, particularly under the situation of endoplasmic reticulum (ER) stress. We examined the effects of hepatocytes conditioned media (CM) from HepG2 treated with ER stress inducers, tunicamycin or thapsigargin
Wenhuo Hu et al.
Blood, 125(14), 2206-2216 (2015-01-31)
Hematopoietic stem cells (HSCs) possess the ability to generate all hematopoietic cell types and to self-renew over long periods, but the mechanisms that regulate their unique properties are incompletely understood. Herein, we show that homozygous deletion of the miR-29a/b-1 bicistron

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service