Merck
  • Enzymatic properties and substrate specificity of a bacterial phosphatidylcholine synthase.
Enzymatic properties and substrate specificity of a bacterial phosphatidylcholine synthase.
The FEBS journal (2014-06-17)
Meriyem Aktas, Stefan Köster, Sarah Kizilirmak, Javier C Casanova, Heidi Betz, Christiane Fritz, Roman Moser, Özkan Yildiz, Franz Narberhaus
ABSTRACT
Phosphatidylcholine (PC) is a rare membrane lipid in bacteria, but is crucial for virulence of the plant pathogen Agrobacterium tumefaciens and various other pathogens. Agrobacterium tumefaciens uses two independent PC biosynthesis pathways. One is dependent on the integral membrane protein PC synthase (Pcs), which catalyzes the conversion of cytidine diphosphate-diacylglycerol (CDP-DAG) and choline to PC, thereby releasing a cytidine monophosphate (CMP). Here, we show that Pcs consists of eight transmembrane segments with its N- and C-termini located in the cytoplasm. A cytoplasmic loop between the second and third membrane helix contains the majority of the conserved amino acids of a CDP-alcohol phosphotransferase motif (DGX2 ARX12 GX3 DX3 D). Using point mutagenesis, we provide evidence for a crucial role of this motif in choline binding and enzyme activity. To study the catalytic features of the enzyme, we established a purification protocol for recombinant Pcs. The enzyme forms stable oligomers and exhibits broad substrate specificity towards choline derivatives. The presence of CDP-DAG and manganese is a prerequisite for cooperative binding of choline. PC formation by Pcs is reversible and proceeds via two successive reactions. In a first choline- and manganese-independent reaction, CDP-DAG is hydrolyzed releasing a CMP molecule. The resulting phosphatidyl intermediate reacts with choline in a second manganese-dependent step to form PC. Pcs and Pcs bind by molecular sieving (1, 2, 3).
Sigma-Aldrich
Acetic acid, 99.5-100.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
1-Propanol, SAJ first grade, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetic acid, SAJ first grade, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetic acid, JIS special grade, ≥99.7%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetic acid solution, 1 M, 1 N
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetic acid, ≥99.7%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetic acid, ≥99.7%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Chloroform, JIS 300, ≥99.0%, for residue analysis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Chloroform, JIS special grade, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Chloroform, SAJ super special grade, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Chloroform, SAJ first grade, ≥99.0%, contains 0.4-0.8% ethanol
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Chloroform, suitable for HPLC
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
1-Propanol, JIS special grade
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
1-Propanol, ≥99%, FG
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
SKU
Pack Size
Availability
Price
Quantity
Supelco
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
1-Propanol, natural, ≥98%, FG
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
1-Propanol, anhydrous, 99.7%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Cytidine, 99%