Skip to Content
Merck
All Photos(3)

Key Documents

279544

Sigma-Aldrich

1-Propanol

anhydrous, 99.7%

Synonym(s):

Propyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2OH
CAS Number:
Molecular Weight:
60.10
Beilstein:
1098242
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
grade:
anhydrous
bp:
97 °C (lit.)
vapor pressure:
10 mmHg ( 147 °C)
14.9 mmHg ( 20 °C)

grade

anhydrous

Quality Level

vapor density

2.1 (vs air)

vapor pressure

10 mmHg ( 147 °C)
14.9 mmHg ( 20 °C)

Assay

99.7%

form

liquid

autoignition temp.

700 °F

expl. lim.

13.7 %

impurities

<0.005% water

evapn. residue

<0.0003%

refractive index

n20/D 1.384 (lit.)

pH

8.5 (20 °C, 200 g/L)

bp

97 °C (lit.)

mp

−127 °C (lit.)

density

0.804 g/mL at 25 °C (lit.)

SMILES string

CCCO

InChI

1S/C3H8O/c1-2-3-4/h4H,2-3H2,1H3

InChI key

BDERNNFJNOPAEC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-Propanol may be used as a starting material in the vapour-phase synthesis of 2-propylphenol and 3-pentanone over CeO2-MgO and CeO2-Fe2O3 catalysts, respectively. It may also be used as a solvent for the palladium catalyzed cross-coupling of boronic acids with aryl halides to form biaryls.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

71.6 °F - closed cup

Flash Point(C)

22 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of unsymmetrical biaryls using a modified Suzuki cross-coupling: 4-biphenylcarboxaldehyde
Bret EH, et al.
Organic Syntheses, 75, 53-53 (1998)
Ortho-selective alkylation of phenol with 1-propanol catalyzed by CeO2-MgO
Sato S, et al
J. Catal., 184(1), 180-188 (1999)
Synthesis of 3-pentanone from 1-propanol over CeO2-Fe2O3 catalysts
Kamimura Y, et al
Applied Catalysis A: General, 252(2), 399-410 (2003)
Mesoporous alumina (I): comparison of synthesis schemes using anionic, cationic, and non-ionic surfactants.
Ray JC, et al.
Microporous and Mesoporous Materials : The Official Journal of the International Zeolite Association, 100(1), 183-190 (2007)
Thijs Pasman et al.
Membranes, 10(11) (2020-11-11)
Polymeric membranes are widely applied in biomedical applications, including in vitro organ models. In such models, they are mostly used as supports on which cells are cultured to create functional tissue units of the desired organ. To this end, the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service