Skip to Content
Merck
All Photos(1)

Key Documents

24-6070

Sigma-Aldrich

1-Propanol

JIS special grade

Synonym(s):

Propyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2OH
CAS Number:
Molecular Weight:
60.10
Beilstein:
1098242
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

grade

JIS special grade

vapor density

2.1 (vs air)

vapor pressure

10 mmHg ( 147 °C)
14.9 mmHg ( 20 °C)

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

700 °F

expl. lim.

13.7 %

availability

available only in Japan

refractive index

n20/D 1.384 (lit.)

pH

8.5 (20 °C, 200 g/L)

bp

97 °C (lit.)

mp

−127 °C (lit.)

density

0.804 g/mL at 25 °C (lit.)

SMILES string

CCCO

InChI

1S/C3H8O/c1-2-3-4/h4H,2-3H2,1H3

InChI key

BDERNNFJNOPAEC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

59.0 °F

Flash Point(C)

15 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dieuwke Sevenster et al.
Science (New York, N.Y.), 339(6121), 830-833 (2013-02-16)
Although reconsolidation opens up new avenues to erase excessive fear memory, subtle boundary conditions put constraints on retrieval-induced plasticity. Reconsolidation may only take place when memory reactivation involves an experience that engages new learning (prediction error). Thus far, it has
Anja K K Rahn et al.
Journal of agricultural and food chemistry, 61(37), 8743-8751 (2013-08-24)
Chloropropanol (CP) esters are part of an emerging group of process-induced toxicants that are considered as potential health hazards particularly in palm oil. Mass spectrometry-based methodologies for identification of CP esters in food are critical in overcoming the challenges associated
Elias N Glaros et al.
The Journal of biological chemistry, 280(26), 24515-24523 (2005-05-14)
Cellular glycosphingolipid (GSL) storage is known to promote cholesterol accumulation. Although physical interactions between GSLs and cholesterol are thought to cause intracellular cholesterol "trapping," it is not known whether cholesterol homeostatic mechanisms are also impaired under these conditions. ApoA-I-mediated cholesterol
David Fernandez Rivas et al.
Ultrasonics sonochemistry, 19(6), 1252-1259 (2012-05-23)
Micromachined pits on a substrate can be used to nucleate and stabilize microbubbles in a liquid exposed to an ultrasonic field. Under suitable conditions, the collapse of these bubbles can result in light emission (sonoluminescence, SL). Hydroxyl radicals (OH()) generated
Takeshi Morita et al.
The journal of physical chemistry. B, 116(24), 7328-7333 (2012-06-01)
We characterized the effects of ethanol (ET) and dimethyl sulfoxide (DMSO) on H(2)O within a limited H(2)O-rich region by the 1-propanol (1P)-probing methodology developed by us earlier. The results are displayed on a two-dimensional map with twin coordinates: one pertaining

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service