Skip to Content
Merck
All Photos(1)

Key Documents

05-3400

Sigma-Aldrich

Chloroform

JIS special grade, ≥99.0%

Synonym(s):

Methylidyne trichloride, Trichloromethane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CHCl3
CAS Number:
Molecular Weight:
119.38
Beilstein:
1731042
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
grade:
JIS special grade
bp:
60.5-61.5 °C (lit.)
vapor pressure:
160 mmHg ( 20 °C)

grade

JIS special grade

vapor density

4.1 (vs air)

vapor pressure

160 mmHg ( 20 °C)

Assay

≥99.0%

form

liquid

availability

available only in Japan

dilution

(for analytical testing)

refractive index

n20/D 1.445 (lit.)

bp

60.5-61.5 °C (lit.)

mp

−63 °C (lit.)

density

1.492 g/mL at 25 °C (lit.)

SMILES string

ClC(Cl)Cl

InChI

1S/CHCl3/c2-1(3)4/h1H

InChI key

HEDRZPFGACZZDS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Accalia Fu et al.
Diabetologia, 58(7), 1513-1522 (2015-04-16)
Precise regulation of insulin secretion by the pancreatic beta cell is essential for the maintenance of glucose homeostasis. Insulin secretory activity is initiated by the stepwise breakdown of ambient glucose to increase cellular ATP via glycolysis and mitochondrial respiration. Knockout
Kuan Sun et al.
Nature communications, 6, 6013-6013 (2015-01-15)
Solution-processed organic photovoltaic cells (OPVs) hold great promise to enable roll-to-roll printing of environmentally friendly, mechanically flexible and cost-effective photovoltaic devices. Nevertheless, many high-performing systems show best power conversion efficiencies (PCEs) with a thin active layer (thickness is ~100 nm) that
Mette Bohl et al.
The American journal of clinical nutrition, 101(4), 870-878 (2015-04-04)
Abdominal obesity and exaggerated postprandial lipemia are independent risk factors for cardiovascular disease (CVD) and mortality, and both are affected by dietary behavior. We investigated whether dietary supplementation with whey protein and medium-chain saturated fatty acids (MC-SFAs) improved postprandial lipid
Todd A Lydic et al.
Methods (San Diego, Calif.), 87, 83-95 (2015-04-25)
There is an increasing recognition of the role that cancer cell derived exosomes play in intercellular signaling upon fusion or uptake with a target cell, including immune system evasion, tumor growth and metastasis. To date, however, although exosomal membrane and
Shuiquan Tang et al.
Philosophical transactions of the Royal Society of London. Series B, Biological sciences, 368(1616), 20120318-20120318 (2013-03-13)
Two novel reductive dehalogenases (RDases) that are highly similar to each other but catalyse distinct dechlorination reactions were identified from Dehalobacter-containing mixed cultures. These two RDases were partially purified from crude protein extracts of anaerobic dechlorinating enrichment cultures using blue

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service