Skip to Content
Merck
All Photos(1)

Key Documents

W292818

Sigma-Aldrich

1-Propanol

≥99%, FG

Synonym(s):

Propyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2OH
CAS Number:
Molecular Weight:
60.10
FEMA Number:
2928
Beilstein:
1098242
EC Number:
Council of Europe no.:
50
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.002
NACRES:
NA.21
grade:
FG
Fragrance grade
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 872/2012

vapor density

2.1 (vs air)

vapor pressure

10 mmHg ( 147 °C)
14.9 mmHg ( 20 °C)

Assay

≥99%

form

liquid

autoignition temp.

700 °F

expl. lim.

13.7 %

refractive index

n20/D 1.384 (lit.)

pH

8.5 (20 °C, 200 g/L)

bp

97 °C (lit.)

mp

−127 °C (lit.)

density

0.804 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

alcohol; musty

SMILES string

CCCO

InChI

1S/C3H8O/c1-2-3-4/h4H,2-3H2,1H3

InChI key

BDERNNFJNOPAEC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Noncontact In Situ Multidiagnostic NMR/Dielectric Spectroscopy: Details advancements in NMR and dielectric spectroscopy techniques, where 1-Propanol is utilized to investigate solvent interactions and molecular dynamics (Morais et al., 2024).

Biochem/physiol Actions

Taste at 25-250 ppm

Other Notes

Natural occurrence: Apple, apricot, banana, beer, gin, honey, pear, sherry, and tea.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

71.6 °F - closed cup

Flash Point(C)

22 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anja K K Rahn et al.
Journal of agricultural and food chemistry, 61(37), 8743-8751 (2013-08-24)
Chloropropanol (CP) esters are part of an emerging group of process-induced toxicants that are considered as potential health hazards particularly in palm oil. Mass spectrometry-based methodologies for identification of CP esters in food are critical in overcoming the challenges associated
Dieuwke Sevenster et al.
Science (New York, N.Y.), 339(6121), 830-833 (2013-02-16)
Although reconsolidation opens up new avenues to erase excessive fear memory, subtle boundary conditions put constraints on retrieval-induced plasticity. Reconsolidation may only take place when memory reactivation involves an experience that engages new learning (prediction error). Thus far, it has
Elias N Glaros et al.
The Journal of biological chemistry, 280(26), 24515-24523 (2005-05-14)
Cellular glycosphingolipid (GSL) storage is known to promote cholesterol accumulation. Although physical interactions between GSLs and cholesterol are thought to cause intracellular cholesterol "trapping," it is not known whether cholesterol homeostatic mechanisms are also impaired under these conditions. ApoA-I-mediated cholesterol
David Fernandez Rivas et al.
Ultrasonics sonochemistry, 19(6), 1252-1259 (2012-05-23)
Micromachined pits on a substrate can be used to nucleate and stabilize microbubbles in a liquid exposed to an ultrasonic field. Under suitable conditions, the collapse of these bubbles can result in light emission (sonoluminescence, SL). Hydroxyl radicals (OH()) generated
Takeshi Morita et al.
The journal of physical chemistry. B, 116(24), 7328-7333 (2012-06-01)
We characterized the effects of ethanol (ET) and dimethyl sulfoxide (DMSO) on H(2)O within a limited H(2)O-rich region by the 1-propanol (1P)-probing methodology developed by us earlier. The results are displayed on a two-dimensional map with twin coordinates: one pertaining

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service