Merck
Plasmacatalytic removal of lead acetate assisted by precipitation.
Chemosphere (2014-01-28)
Nabila Haddou, Mouffok Redouane Ghezzar, Fatiha Abdelmalek, Stéphanie Ognier, Marc Martel, Ahmed Addou
ABSTRACT
The Gliding Arc Discharge (GAD) is an efficient non-thermal plasma technique able to degrade organic compounds dispersed in water at atmospheric pressure. The degradation of the organometallic lead acetate (PbAc) in aqueous solution was performed by two distinct plasmageneous processes: GAD and GAD/TiO2. The global oxidation of the organic matter was followed by Chemical Oxygen Demand (COD) and the mineralization was determined by the Total Organic Carbon (TOC). The Pb(2+) ions released during the degradation process were measured by Atomic Absorption Spectroscopy (AAS). For 2h of GAD treatment, the degradation rate of PbAc (10mM) reached 83% and for the same duration of GAD/TiO2 process ([TiO2]=1gL(-1)), it reached 93%. The release of Pb(2+) ions in the solution was respectively of 95% and 57% for GAD and GAD/TiO2 processes. The released Pb(2+) ions were removed by precipitation process in a basic medium at pH=11.1. A reaction mechanism was proposed to explain the PbAc molecule degradation and the Pb(2+) elimination.
Sigma-Aldrich
Titanium(IV) oxide, JIS special grade, ≥99.0%
Sigma-Aldrich
Titanium(IV) oxide, contains 1% Mn as dopant, nanopowder, <100 nm particle size (BET), ≥97%
Sigma-Aldrich
Titanium(IV) oxide, nanowires, diam. × L ~10 nm × 10 μm
Sigma-Aldrich
Titanium(IV) oxide, nanowires, diam. × L ~100 nm × 10 μm
Sigma-Aldrich
Titanium(IV) oxide, anatase, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, nanopowder, 21 nm primary particle size (TEM), ≥99.5% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, anatase, powder, 99.8% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, anatase, nanopowder, <25 nm particle size, 99.7% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, puriss., meets analytical specification of Ph. Eur., BP, USP, 99-100.5%
Sigma-Aldrich
Titanium(IV) oxide, ReagentPlus®, ≥99%
Sigma-Aldrich
Lead(II) acetate trihydrate, JIS special grade, ≥99.5%
Sigma-Aldrich
Titanium(IV) oxide, rutile, <001>, (single crystal substrate), ≥99.9% trace metals basis, L × W × thickness 10 mm × 10 mm × 0.5 mm
Sigma-Aldrich
Lead(II) acetate trihydrate, 99.999% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, 99.995% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, ≥99.98% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, mixture of rutile and anatase, nanoparticles, <150 nm particle size (volume distribution, DLS), dispersion, 40 wt. % in H2O, 99.5% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, mixture of rutile and anatase, nanopowder, <100 nm particle size (BET), 99.5% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, powder, <5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, nanopowder, <100 nm particle size, 99.5% trace metals basis
Sigma-Aldrich
Lead(II) acetate trihydrate, ACS reagent, ≥99%
Sigma-Aldrich
Lead(II) acetate trihydrate, ≥99.99% trace metals basis
Sigma-Aldrich
Lead(II) acetate trihydrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.5-102.0%