Skip to Content
Merck
All Photos(3)

Documents

232033

Sigma-Aldrich

Titanium(IV) oxide, anatase

powder, 99.8% trace metals basis

Synonym(s):

Titanium dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TiO2
CAS Number:
Molecular Weight:
79.87
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

Assay

99.8% trace metals basis

form

powder

mp

1825 °C (lit.)

density

3.9 g/mL at 25 °C (lit.)

SMILES string

O=[Ti]=O

InChI

1S/2O.Ti

InChI key

GWEVSGVZZGPLCZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Titanium(IV) oxide is a chemically inert and non-toxic semiconductormaterial widely used in the field of photovoltaics and environmentalremediation. Thephotocatalytic activity of anatase is superior tothat of other polymorphs of titanium oxide.

Application

Titanium(IV) oxide can be used as a precursor:
  • To synthesize TiO2/ZnO- calcium alginate photocatalyst for removal of copper from aqueous solutions.
  • To fabricate implantable composite bioelectrode for biosensing applications.
It can also be used as a nanoparticle growth initiator for the anodic modification of microbial fuel cells(MFC) to enhance their performance.

Features and Benefits

  • Higher electron mobility
  • Low dielectric constant
  • Lower ability to adsorb oxygen
  • Higher Fermi level
  • High degree of hydroxylation
  • Superior biocompatibility
  • Corrosion resistance

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qi Fang et al.
Biomedical optics express, 10(8), 4034-4049 (2019-08-28)
Optical coherence elastography (OCE) has been proposed for a range of clinical applications. However, the majority of these studies have been performed using bulky, lab-based imaging systems. A compact, handheld imaging probe would accelerate clinical translation, however, to date, this
Sanjay Thorat et al.
Dental materials journal, 31(4), 635-644 (2012-08-07)
A photo-polymerizable Bisphenol-A diglycidylether methacrylate resin was characterized by Fourier transform infrared spectroscopy after its irradiation under different conditions to identify the best curing. Bonding-agent free composites with particles of ball-milled glass, silica and titania at loading of 10 and
Zhiyuan Liu et al.
Environmental science and pollution research international, 29(16), 23352-23360 (2021-11-24)
The occurrence of titanium dioxide nanoparticle (TNP), an emerging contaminant, in Taihu Lake of China was investigated. Ti was present at a concentration of 224 ± 59 μg/L in the water samples collected from a water source in east Taihu
Brooke Krajancich et al.
Biomedical optics express, 10(1), 226-241 (2019-02-19)
Handheld imaging probes are needed to extend the clinical translation of optical elastography to in vivo applications, yet such probes have received little attention. In this paper, we present the first demonstration of optical palpation using a handheld probe. Optical
Qi Fang et al.
Biomedical optics express, 10(8), 4034-4049 (2019-08-28)
Optical coherence elastography (OCE) has been proposed for a range of clinical applications. However, the majority of these studies have been performed using bulky, lab-based imaging systems. A compact, handheld imaging probe would accelerate clinical translation, however, to date, this

Articles

TiO2 exhibits wide band gap semiconductor and memristor properties electronically, with high opacity and UV absorbance optically.

TiO2 exhibits wide band gap semiconductor and memristor properties electronically, with high opacity and UV absorbance optically.

TiO2 exhibits wide band gap semiconductor and memristor properties electronically, with high opacity and UV absorbance optically.

TiO2 exhibits wide band gap semiconductor and memristor properties electronically, with high opacity and UV absorbance optically.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service