Skip to Content
Merck
All Photos(1)

Documents

677469

Sigma-Aldrich

Titanium(IV) oxide

contains 1% Mn as dopant, nanopowder, <100 nm particle size (BET), ≥97%

Synonym(s):

Titania, Titanium dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TiO2
CAS Number:
Molecular Weight:
79.87
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

≥97%

form

nanopowder

contains

1% Mn as dopant

reaction suitability

reagent type: catalyst
core: titanium

surface area

>14.0 m2/g

particle size

<100 nm (BET)

mp

>350 °C (lit.)

density

4.26 g/mL at 25 °C (lit.)

SMILES string

O=[Ti]=O

InChI

1S/2O.Ti

InChI key

GWEVSGVZZGPLCZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Titanium dioxide nanopowder was used to generate light scattering tissue phantoms. Its effect on the development of Xenopus laevis was also investigated. Titania nanopowder may be used for coating as well.

Legal Information

Product of Engi-Mat Co.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of ZnO Aggregates and Their Application in Dye-sensitized Solar Cells
Qifeng Z, et al.
Material Matters, 5(2), 32-32 (2010)
Renato Bacchetta et al.
Nanotoxicology, 6(4), 381-398 (2011-05-18)
The teratogenic potential of commercially available copper oxide (CuO), titanium dioxide (TiO2) and zinc oxide (ZnO) nanoparticles (NPs) was evaluated using the standardized FETAX test. After characterization of NP suspensions by TEM, DLS and AAS, histopathological screening and advanced confocal
Joshua K Meisner et al.
Microcirculation (New York, N.Y. : 1994), 19(7), 619-631 (2012-05-18)
1) To develop and validate laser speckle flowmetry (LSF) as a quantitative tool for individual microvessel hemodynamics in large networks. 2) To use LSF to determine if structural differences in the dorsal skinfold microcirculation (DSFWC) of C57BL/6 and BALB/c mice
Nabila Haddou et al.
Chemosphere, 107, 304-310 (2014-01-28)
The Gliding Arc Discharge (GAD) is an efficient non-thermal plasma technique able to degrade organic compounds dispersed in water at atmospheric pressure. The degradation of the organometallic lead acetate (PbAc) in aqueous solution was performed by two distinct plasmageneous processes:
D Minetto et al.
Environment international, 66, 18-27 (2014-02-11)
The innovative properties of nanomaterials make them suitable for various applications in many fields. In particular, TiO2 nanoparticles (nTiO2) are widely used in paints, in cosmetics and in sunscreens that are products accessible to the mass market. Despite the great

Articles

Operation principle and market dominance of single crystalline silicon solar cells.

Operation principle and market dominance of single crystalline silicon solar cells.

Operation principle and market dominance of single crystalline silicon solar cells.

Operation principle and market dominance of single crystalline silicon solar cells.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service