Skip to Content
Merck
All Photos(1)

Key Documents

32307

Sigma-Aldrich

Lead(II) acetate trihydrate

puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.5-102.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pb(CH3CO2)2 · 3H2O
CAS Number:
Molecular Weight:
379.33
Beilstein:
3730298
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
Assay:
99.5-102.0%
form:
solid

grade

ACS reagent
puriss. p.a.

Quality Level

Agency

USP/NF
reag. ISO
reag. Ph. Eur.

Assay

99.5-102.0%

form

solid

impurities

≤0.005% insoluble in acetic acid

mp

75 °C (dec.) (lit.)

anion traces

chloride (Cl-): ≤5 mg/kg
nitrate (NO3-): ≤10 mg/kg

cation traces

Ca: ≤50 mg/kg
Cd: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤10 mg/kg
K: ≤50 mg/kg
Mg: ≤50 mg/kg
Na: ≤50 mg/kg
Zn: ≤5 mg/kg

SMILES string

O.O.O.CC(=O)O[PbH2]OC(C)=O

InChI

1S/2C2H4O2.3H2O.Pb/c2*1-2(3)4;;;;/h2*1H3,(H,3,4);3*1H2;/q;;;;;+2/p-2

InChI key

MCEUZMYFCCOOQO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Lead(II) acetate is a white crystalline compound. It is widely employed in cosmetic products. Lead acetate paper is employed for the detection of poisonous gas, hydrogen sulfide. It reacts with 5-ethynyl-1,3-benzenedicarboxylic acid (H2ebdc) to afford a 1D ladder network, [Pb(ebdc)(MeOH)]2·H2O.

Application

Lead(II) acetate trihydrate has been used in the preparation of PbSe-CdSe-PbSe nanodumbbells.

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Lact. - Repr. 1A - STOT RE 1

Target Organs

Central nervous system,Blood,Immune system,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Hazardous Chemicals: Safety Management and Global Regulations, 244-244 (2013)
Carbone L, et al.
Journal of Materials Chemistry, 16(40), 3952-3956 (2006)
Jane V Knichal et al.
Chemical communications (Cambridge, England), 50(92), 14436-14439 (2014-10-11)
5-Ethynyl-1,3-benzenedicarboxylic acid (H2ebdc) reacted with lead(II) acetate trihydrate yields a 1D ladder network, [Pb(ebdc)(MeOH)]2·H2O (1). Removing crystals of 1 from the mother liquor results in a facile single crystal to single crystal transition, yielding 2D [Pb(ebdc)] net (2) with a
Asit Rai et al.
Toxicology mechanisms and methods, 23(2), 99-107 (2012-08-21)
Exposure to a mixture of As, Pb and Cd induces apoptosis and morphological alterations in the cortical astrocytes of rat brain. The levels of the glial fibrillary acidic protein (GFAP) undergo a reduction. The GFAP exists in several isoforms, viz.
I Baranowska-Bosiacka et al.
Toxicology, 303, 187-200 (2012-11-14)
The aim of this paper is to examine if pre- and neonatal exposure to lead (Pb) may intensify or inhibit apoptosis or necroptosis in the developing rat brain. Pregnant experimental females received 0.1% lead acetate (PbAc) in drinking water from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service