Skip to Content
Merck
All Photos(5)

Documents

637254

Sigma-Aldrich

Titanium(IV) oxide, anatase

nanopowder, <25 nm particle size, 99.7% trace metals basis

Synonym(s):

TiO2 nanopowder, Titanium dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TiO2
CAS Number:
Molecular Weight:
79.87
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

99.7% trace metals basis

form

nanopowder

surface area

45-55 m2/g

particle size

<25 nm

mp

1825 °C (lit.)

density

3.9 g/mL at 25 °C (lit.)

bulk density

0.04‑0.06 g/mL

application(s)

battery manufacturing

SMILES string

O=[Ti]=O

InChI

1S/2O.Ti

InChI key

GWEVSGVZZGPLCZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Our titanium(IV) oxide, anatase nanopowder is a fine white powder composed of titanium dioxide nanoparticles with a particle size less than 25 nm. Anatase is a metastable polymorph of TiO2, which is less hard and less dense than the rutile polymorph. Optically, anatase nanopowder has a lower refractive index, absorbs less UV light, and exhibits greater photocatalytic activity than the rutile polymorph. Consequently, anatase is often preferred in applications where photocatalytic activity is desired, such as self-cleaning surfaces and solar cells.

Application

Titania paste may be used as a transparent coating for self cleaning glass.Low optical scattering titania-acrylate nanocomposites have been reported. Metal contacts in solar cells based on titanium dioxide and di-(isothiocyanate)-bis-(2,2′-bipyridyl-4,4′-dicarboxylate)ruthenium(II) have been studied.

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mojca Pavlin et al.
International journal of molecular sciences, 23(11) (2022-06-11)
Alongside physiochemical properties (PCP), it has been suggested that the protein corona of nanoparticles (NPs) plays a crucial role in the response of immune cells to NPs. However, due to the great variety of NPs, target cells, and exposure protocols
Saher Fatima et al.
Pharmaceutics, 13(10) (2021-10-24)
Multi-drug resistant (MDR) bacterial cells embedded in biofilm matrices can lead to the development of chronic cariogenesis. Here, we isolated and identified three Gram-positive MDR oral cocci, (1) SJM-04, (2) SJM-38, and (3) SJM-65, and characterized them morphologically, biochemically, and
TiO2-Acrylate Nanocomposites Elaborated by UV-Curing with Tunable Properties.
Burunkova JA, et al.
Solid State Phenomena, 200, 173-177 (2013)
Preparation of small particles of titanium (IV) oxide
Mohs CR and Klabunde KJ
Nanophase Materials, 260, 121-124 (1994)
Cell cycle synchronization reveals greater G2/M-phase accumulation of lung epithelial cells exposed to titanium dioxide nanoparticles
Medina-Reyes, et al.
Environmental Science and Pollution Research International, 1-7 (2014)

Articles

Dye-sensitized solar cells directly convert sunlight to electricity

Dye-sensitized solar cells (DSSCs) attract attention for high performance and potential low-cost production in solar energy.

Dye-sensitized solar cells (DSSCs) attract attention for high performance and potential low-cost production in solar energy.

Dye-sensitized solar cells (DSSCs) attract attention for high performance and potential low-cost production in solar energy.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service