Merck
  • Decreased serum levels of D-serine in patients with schizophrenia: evidence in support of the N-methyl-D-aspartate receptor hypofunction hypothesis of schizophrenia.
Decreased serum levels of D-serine in patients with schizophrenia: evidence in support of the N-methyl-D-aspartate receptor hypofunction hypothesis of schizophrenia.
Archives of general psychiatry (2003-06-11)
Kenji Hashimoto, Takeshi Fukushima, Eiji Shimizu, Naoya Komatsu, Hiroyuki Watanabe, Naoyuki Shinoda, Michiko Nakazato, Chikara Kumakiri, Shin-ichi Okada, Hisanori Hasegawa, Kazuhiro Imai, Masaomi Iyo
ABSTRACT
The hypofunction of the N-methyl-D-aspartate (NMDA) subtype of glutamate receptors has been implicated in the pathophysiology of schizophrenia. Several lines of evidence suggest that D-serine may function as an endogenous agonist of the glycine site of the NMDA receptor. The aim of this study was to examine whether serum levels of D- and L-serine in patients with schizophrenia are different from those of healthy controls. Forty-two patients with schizophrenia and 42 age- and sex-matched healthy controls were enrolled in this study. Symptoms were assessed using the Brief Psychiatric Rating Scale. Serum levels of total serine and D- and L-serine were measured by high-performance liquid chromatography. Serum levels of D-serine in the patients with schizophrenia were significantly (z = -3.30, P =.001) lower than those of healthy controls. In contrast, serum levels of total (D and L) serine (z = -2.40, P =.02) and L-serine (z = -2.49, P =.01) in the schizophrenic patients were significantly higher than those of controls. In addition, the percentage of D-serine in the total serine in the schizophrenic patients was significantly (z = -4.78, P<.001) lower than that of controls, suggesting that the activity of serine racemase, an enzyme catalyzing the formation of D-serine from L-serine, may have been reduced in the schizophrenic patients. Reduced levels of D-serine may play a role in the pathophysiology of schizophrenia, and serum D- and L-serine levels might provide a measurable biological marker for schizophrenia.
Product Description
Supelco
L-Threonine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Tyrosine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Alanine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Aspartic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Alanine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
SAFC
L-Glutamine Solution 200 mM, 29.23 mg/mL in saline, solution, suitable for cell culture
Supelco
L-Tyrosine, Pharmaceutical Secondary Standard; Certified Reference Material
Alanine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Glycine, SAJ special grade, ≥99.0%
SAFC
L-Aspartic acid
Sigma-Aldrich
L-Aspartic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
L-Tyrosine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
L-Threonine, BioXtra, ≥99.5% (NT)
Sigma-Aldrich
L-Tyrosine, FG
Sigma-Aldrich
4-Hydroxyphenylacetic acid, 98%
Sigma-Aldrich
L-Alanine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Alanine, ≥99%
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
L-Glutamine solution, 200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine solution, 200 mM, Hybri-Max, sterile-filtered, suitable for hybridoma
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
L-Tyrosine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Aspartic acid, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
L-Aspartic acid, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
L-Threonine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Threonine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%