Skip to Content
Merck
All Photos(1)

Key Documents

W381829

Sigma-Aldrich

L-Alanine

≥99%

Synonym(s):

(S)-2-Aminopropionic acid, L-α-Aminopropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H7NO2
CAS Number:
Molecular Weight:
89.09
FEMA Number:
3818
Beilstein:
1720248
EC Number:
MDL number:
UNSPSC Code:
12164502
eCl@ss:
32160406
PubChem Substance ID:
Flavis number:
17.002
NACRES:
NA.21
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Assay

≥99%

form

chunks
crystalline powder
powder

optical activity

[α]20/D +13.5 to +15.5°, c = 10 in 6 M HCl

solubility

H2O: soluble 89.1 g/L at 20 °C (completely)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

sweet

SMILES string

C[C@H](N)C(O)=O

InChI

1S/C3H7NO2/c1-2(4)3(5)6/h2H,4H2,1H3,(H,5,6)/t2-/m0/s1

InChI key

QNAYBMKLOCPYGJ-REOHCLBHSA-N

Gene Information

human ... CA1(759) , CA2(760)

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • Exposure to synthesized tribromobisphenol A and critical effects: Metabolic pathways, disease signature, and benchmark dose derivation.: This study provides insight into the metabolic pathways and disease signatures associated with exposure to synthesized tribromobisphenol A, emphasizing the critical role of L-alanine in mediating these effects (Kuang et al., 2024).

  • Prenylated indole diketopiperazine alkaloids as phosphatase inhibitors from the marine-derived fungus Talaromyces purpureogenus.: Identifies new prenylated indole diketopiperazine alkaloids from Talaromyces purpureogenus that act as potent phosphatase inhibitors, with L-alanine being key to their structure and bioactivity, offering potential for drug development (Liang et al., 2024).


Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Marta Guembe-García et al.
Polymers, 12(6) (2020-06-04)
We anchored a colourimetric probe, comprising a complex containing copper (Cu(II)) and a dye, to a polymer matrix obtaining film-shaped chemosensors with induced selectivity toward glycine. This sensory material is exploited in the selectivity detection of glycine in complex mixtures
Lillian L Siu et al.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology, 31(19), 2477-2484 (2013-05-22)
The antiepidermal growth factor receptor monoclonal antibody cetuximab has improved survival in patients with metastatic, chemotherapy-refractory, wild-type K-RAS colorectal cancer. The addition of brivanib, a tyrosine kinase inhibitor targeting vascular endothelial growth factor receptor and fibroblast growth factor receptor, to
J M Rhoads et al.
Journal of pediatric gastroenterology and nutrition, 9(2), 225-231 (1989-08-01)
We studied sodium-dependent uptake of L-alanine into small intestinal brush border membrane vesicles (BBMV) isolated from piglets 40 h after infection with transmissible gastroenteritis (TGE) virus. Vesicles from TGE-infected pigs and uninfected litter-mate controls showed comparable degrees of enrichment and
Mom Das et al.
Nucleic acids research, 42(6), 3943-3953 (2013-12-29)
Errors in protein synthesis due to mispairing of amino acids with tRNAs jeopardize cell viability. Several checkpoints to prevent formation of Ala- and Cys-tRNA(Pro) have been described, including the Ala-specific editing domain (INS) of most bacterial prolyl-tRNA synthetases (ProRSs) and
Rachelle S Doody et al.
The New England journal of medicine, 369(4), 341-350 (2013-07-26)
Alzheimer's disease is characterized by the presence of cortical amyloid-beta (Aβ) protein plaques, which result from the sequential action of β-secretase and γ-secretase on amyloid precursor protein. Semagacestat is a small-molecule γ-secretase inhibitor that was developed as a potential treatment

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service