Skip to Content
Merck
All Photos(4)

Key Documents

T8566

Sigma-Aldrich

L-Tyrosine

99.0-101.0%, suitable for cell culture, BioXtra, non-animal source

Synonym(s):

(S)-2-Amino-3-(4-hydroxyphenyl)propionic acid, 3-(4-Hydroxyphenyl)-L-alanine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(HO)C6H4CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
181.19
Beilstein:
392441
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

Product Name

L-Tyrosine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 99.0-101.0%

biological source

non-animal source

Agency

USP/NF
meets EP testing specifications
meets USP testing specifications

product line

BioXtra

Assay

99.0-101.0%

form

powder

optical activity

[α]25/D 11.2 to 9.8°, c = 5 in 1 M HCl (Specific Rotation 6.1 - 7.8 °)

shelf life

limited shelf life, expiry date on the label

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

color

white to off-white

mp

>300 °C (dec.) (lit.)

solubility

1 M HCl: 25 mg/mL

cation traces

Fe: ≤10 ppm
NH4+: ≤0.02%

application(s)

pharmaceutical (small molecule)

functional group

(phenol)

storage temp.

room temp

SMILES string

N[C@@H](Cc1ccc(O)cc1)C(O)=O

InChI

1S/C9H11NO3/c10-8(9(12)13)5-6-1-3-7(11)4-2-6/h1-4,8,11H,5,10H2,(H,12,13)/t8-/m0/s1

InChI key

OUYCCCASQSFEME-QMMMGPOBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

L-Tyrosine, an essential amino acid, plays a vital role in biochemical and cell culture research. In biochemical studies, L-Tyrosine is significant as a precursor to neurotransmitters like dopamine, norepinephrine, and epinephrine, which influence cellular communication and play a role in mood, attention, and energy levels. It also contributes to the synthesis of thyroid hormones, regulating metabolic processes. L-Tyrosine is employed to support cell growth and protein synthesis, facilitating experiments on cell maintenance and repair, making it a valuable tool in biochemical and cell culture investigations.

Application

L-Tyrosine has been used as a supplement in Dulbecco′s modified Eagle′s medium for culturing SY5Y neuroblastoma cells for stable isotope labeling of amino acids on cell culture (SILAC).

Biochem/physiol Actions

L-Tyrosine consists of a polar side chain and is a non-essential amino acid. It is utilized by cells to synthesize proteins that are involved in signal transduction. L-Tyrosine acts as a receiver of phosphate groups that are transferred by kinases.

Features and Benefits

  • Suitable for Cell Culture and Biochemical research
  • High-quality compound suitable for multiple research applications

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Amino acid precursor of dopamine and other catecholamines
For additional information on our range of Biochemicals, please complete this form.

comparable product

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemistry, An Introduction to General, Organic, and Biological Chemistry (2016)
Altering O-Linked ?-N-Acetylglucosamine Cycling Disrupts Mitochondrial Function
Ee Phie Tan
The Journal of Biological Chemistry, 14719-14730 (2014)
John W Hill
Chemistry for Changing Times (2016)
Pin-Chao Liao et al.
STAR protocols, 1(3), 100160-100160 (2020-12-31)
The redox state of mitochondria is one indicator of the functional state of the organelles. Mitochondria are also the primary endogenous source of reactive oxygen species (ROS). Therefore, the redox state of the organelles also reflects their function in ROS
Verena Rahm et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 55(4), 546-550 (2014-02-26)
Low-grade gliomas (LGGs) may harbor malignant foci, which are characterized by increased tumor cellularity and angiogenesis. We used diffusion-weighted MR imaging (apparent diffusion coefficient [ADC]) and PET with the amino acid O-(2-(18)F-fluorethyl)-L-tyrosine ((18)F-FET) to search for focal changes of diffusion

Articles

Learn more about the amino acid L-Tyrosine and explore how it is used in cell culture media like MEM, DMEM, and RPMI. We offer a comprehensive portfolio of amino acid cell culture supplements and reagents.

Learn more about the amino acid L-Tyrosine and explore how it is used in cell culture media like MEM, DMEM, and RPMI. We offer a comprehensive portfolio of amino acid cell culture supplements and reagents.

Learn more about the amino acid L-Tyrosine and explore how it is used in cell culture media like MEM, DMEM, and RPMI. We offer a comprehensive portfolio of amino acid cell culture supplements and reagents.

Learn more about the amino acid L-Tyrosine and explore how it is used in cell culture media like MEM, DMEM, and RPMI. We offer a comprehensive portfolio of amino acid cell culture supplements and reagents.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service