Skip to Content
Merck
All Photos(1)

Documents

1437009

USP

Methylprednisolone hemisuccinate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

11β,17,21-Trihydroxy-6α-methylpregna-1,4-diene-3,20-dione 21-succinate, Methylprednisolone hydrogen succinate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H34O8
CAS Number:
Molecular Weight:
474.54
UNSPSC Code:
41116107
NACRES:
NA.24

API family

methylprednisolone

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

InChI

1S/C26H34O8.Na/c1-14-10-16-17-7-9-26(33,20(29)13-34-22(32)5-4-21(30)31)25(17,3)12-19(28)23(16)24(2)8-6-15(27)11-18(14)24;/h6,8,11,14,16-17,19,23,28,33H,4-5,7,9-10,12-13H2,1-3H3,(H,30,31);/q;+1/p-1/t14-,16-,17-,19-,23+,24-,25-,26-;/m0./s1

InChI key

FQISKWAFAHGMGT-SGJOWKDISA-M

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Methylprednisolone hemisuccinate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Methylprednisolone Sodium Succinate
  • Methylprednisolone Sodium Succinate for Injection

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B - STOT RE 2

Target Organs

Adrenal gland,Immune system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Severe atopic dermatitis accompanied by autoimmune retinopathy.
Takashi Nomura et al.
European journal of dermatology : EJD, 23(2), 263-264 (2013-04-17)
Kai-feng Wang et al.
Zhonghua wai ke za zhi [Chinese journal of surgery], 51(5), 426-431 (2013-08-21)
To investigate the effects and apoptosis of intrathecal injection of Methylprednisolone Sodium Succinate (MPss) for acute spinal cord injury (SCI) in New Zealand rabbits. Seventy-two healthy New Zealand rabbits were used for the procedure and were randomly divided into two
Long-term selective IgE-mediated hypersensitivity to hydrocortisone sodium succinate.
G F Calogiuri et al.
Allergologia et immunopathologia, 41(3), 206-208 (2012-09-25)
A patient experiencing anaphylaxis and urticaria, in some cases preceded by ingestion of red meat.
Phillip Lieberman et al.
The journal of allergy and clinical immunology. In practice, 1(1), 107-108 (2013-11-16)
Claudia Druschel et al.
Spine, 38(11), E669-E677 (2013-03-01)
Written mail-out survey. To determine current practice in high-dose methylprednisolone succinate (MPSS) administration for treatment of acute spinal cord injury (SCI) in Germany. Reanalysis of the National Acute Spinal Cord Injury Studies (NASCIS) resulted in criticism of the use of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service