Skip to Content
Merck
All Photos(1)

Documents

M0639

Sigma-Aldrich

6α-Methylprednisolone

≥98%

Synonym(s):

11β,17α,21-Trihydroxy-6α-methyl-1,4-pregnadiene-3,20-dione, 6α-Methyl-11β,17α,21-trihydroxy-1,4-pregnadiene-3,20-dione, Medrol, Medrone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H30O5
CAS Number:
Molecular Weight:
374.47
Beilstein:
2340300
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic

sterility

non-sterile

Assay

≥98%

form

powder

solubility

chloroform: methanol (1:1): 50 mg/mL, clear, colorless to faintly yellow

application(s)

cell analysis

shipped in

ambient

storage temp.

room temp

SMILES string

[H][C@@]12C[C@H](C)C3=CC(=O)C=C[C@]3(C)[C@@]1([H])[C@@H](O)C[C@@]4(C)[C@@]2([H])CC[C@]4(O)C(=O)CO

InChI

1S/C22H30O5/c1-12-8-14-15-5-7-22(27,18(26)11-23)21(15,3)10-17(25)19(14)20(2)6-4-13(24)9-16(12)20/h4,6,9,12,14-15,17,19,23,25,27H,5,7-8,10-11H2,1-3H3/t12-,14-,15-,17-,19+,20-,21-,22-/m0/s1

InChI key

VHRSUDSXCMQTMA-PJHHCJLFSA-N

Gene Information

human ... NR3C1(2908)
rat ... Nr3c1(24413)

Looking for similar products? Visit Product Comparison Guide

Application

6α-Methylprednisolone has been used:
  • to stimulate RPMI 8226.1 cells in order to govern the expression of human interleukin-10 (IL-10) gene
  • to induce depletion of CD14+ CD16+ monocytes
  • to study its effect on behavioral and sensory afferent hypersensitivity

Biochem/physiol Actions

Methylprednisolone is a glucocorticoid having anti-inflammatory and immunosuppressive actions. It is useful in treating skin diseases, allergies, multiple sclerosis, asthma, rheumatic disorders, chronic obstructive pulmonary disease, croup and cancer.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B - STOT RE 2

Target Organs

Adrenal gland,Immune system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ling Ye et al.
Anesthesiology, 121(3), 632-643 (2014-05-02)
Localized inflammation of lumbar dorsal root ganglia (DRG) may contribute to low back pain. Local injections of corticosteroids used for low back pain are sometimes ineffective. Many corticosteroids activate not only the target glucocorticoid receptor (GR) but also the mineralocorticoid
Mechanism of glucocorticoid-induced depletion of human CD14+ CD16+ monocytes
Dayyani F, et al.
Journal of Leukocyte Biology, 74(1), 33-39 (2003)
Role of STAT3 in glucocorticoid-induced expression of the human IL-10 gene
Unterberger C, et al.
Molecular Immunology, 45(11), 3230-3237 (2008)
Blocking the mineralocorticoid receptor improves effectiveness of steroid treatment for low back pain in rats
Ye L, et al.
Anesthesiology, 121(3), 632-643 (2014)
Formulation development and evaluation of Methylprednisolone dispersible tablets
Nandhini J and Rajalakshmi AN
Asian Journal of Pharmacy and Pharmacology, 4(4), 514-521 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service