Skip to Content
Merck
All Photos(1)

Key Documents

M1755000

Methylprednisolone acetate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

11β,17,21-Trihydroxy-6α-methylpregna-1,4-diene-3,20-dione 21-acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H32O6
CAS Number:
Molecular Weight:
416.51
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

methylprednisolone

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O[C@]1([C@@]2([C@H]([C@H]3[C@@H]([C@@]4(C(=CC(=O)C=C4)[C@H](C3)C)C)[C@H](C2)O)CC1)C)C(=O)COC(=O)C

InChI

1S/C24H32O6/c1-13-9-16-17-6-8-24(29,20(28)12-30-14(2)25)23(17,4)11-19(27)21(16)22(3)7-5-15(26)10-18(13)22/h5,7,10,13,16-17,19,21,27,29H,6,8-9,11-12H2,1-4H3/t13-,16-,17-,19-,21+,22-,23-,24-/m0/s1

InChI key

PLBHSZGDDKCEHR-LFYFAGGJSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Methylprednisolone acetate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D A Nelson
Archives of neurology, 45(7), 804-806 (1988-07-01)
Clinical trials first began in 1960 with methylprednisolone acetate (Depo-Medrol) administered intrathecally, in an attempt to treat both disk disease and multiple sclerosis. After a few reports of salubrious results, there began an outpouring of contradictory data, which continues in
Jennifer L Lyons et al.
Journal of clinical microbiology, 51(4), 1285-1287 (2013-02-01)
This case series highlights our experience with use of the Fungitell assay for quantifying (1,3)-β-d-glucan in cerebrospinal fluid during the current U.S. outbreak of fungal meningitis related to contaminated methylprednisolone acetate. This test may prove a useful adjunct in diagnosis
Kirk Kleinfeld et al.
JAMA neurology, 70(9), 1173-1176 (2013-07-24)
Fungal meningitis due to injections of contaminated methylprednisolone acetate can present with vascular sequelae in immunocompetent individuals. This is particularly germane to neurologists because better recognition of the clinical characteristics of patients with fungal meningitis and ischemic stroke will provide
Thomas M Kerkering et al.
Annals of internal medicine, 158(3), 154-161 (2012-11-28)
Chinese translation Administration of epidural steroid injections (ESIs) with contaminated methylprednisolone resulted in an outbreak of fungal meningitis in many locations in the United States. To characterize early clinical findings and initial response to treatment. Case series with standardized observation
Peter G Pappas
Current opinion in infectious diseases, 26(6), 545-550 (2013-10-25)
The recent outbreak of fungal meningitis related to contaminated methylprednisolone acetate injections represents an important cause of morbidity and continues to be a significant public health problem in the United States. As of August 2013, there have been 749 cases

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service