Skip to Content
Merck
All Photos(1)

Key Documents

PHR1717

Supelco

Methylprednisolone

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

6α-Methylprednisolone, 11β,17α,21-Trihydroxy-6α-methyl-1,4-pregnadiene-3,20-dione, 6α-Methyl-11β,17α,21-trihydroxy-1,4-pregnadiene-3,20-dione, Medrol, Medrone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H30O5
CAS Number:
Molecular Weight:
374.47
Beilstein:
2340300
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 248
traceable to Ph. Eur. M1750000
traceable to USP 1435003

API family

methylprednisolone

CofA

current certificate can be downloaded

packaging

pkg of 500 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

[H][C@@]12C[C@H](C)C3=CC(=O)C=C[C@]3(C)[C@@]1([H])[C@@H](O)C[C@@]4(C)[C@@]2([H])CC[C@]4(O)C(=O)CO

InChI

1S/C22H30O5/c1-12-8-14-15-5-7-22(27,18(26)11-23)21(15,3)10-17(25)19(14)20(2)6-4-13(24)9-16(12)20/h4,6,9,12,14-15,17,19,23,25,27H,5,7-8,10-11H2,1-3H3/t12-,14-,15-,17-,19+,20-,21-,22-/m0/s1

InChI key

VHRSUDSXCMQTMA-PJHHCJLFSA-N

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Methylprednisolone is a synthetic corticosteroid drug which is employed for the management of inflammatory diseases. It is used to relieve the inflammations caused by psoriasis, eczema, arthritis, asthma and rhinitis. It is also widely employed for the treatment of acute lung injury caused by acid aspiration.

Application

Methylprednisolone may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations and counterfeit cosmetic products by chromatography and electroanalytical techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAB8710 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B - STOT RE 2

Target Organs

Adrenal gland,Immune system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Methylprednisolone
USP42-NF37
United States Pharmacopeia/National Formulary, 44(3), 2859-2859 (2013)
LC-MS method for the simultaneous determination of six glucocorticoids in pharmaceutical formulations and counterfeit cosmetic products
Fiori J and Andrisano V
Journal of Pharmaceutical and Biomedical Analysis, 91, 185-192 (2014)
Differential pulse voltammetric determination of methylprednisolone in pharmaceuticals and human biological fluids
Goyal RN, et al.
Analytica Chimica Acta, 605(1), 34-40 (2007)
Simultaneous determination of cortisol, dexamethasone, methylprednisolone, prednisone, prednisolone, mycophenolic acid and mycophenolic acid glucuronide in human plasma utilizing liquid chromatography-tandem mass spectrometry
DiFrancesco R, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 859(1), 42-51 (2007)
Determination of methylprednisolone acetate in biological fluids at gold nanoparticles modified ITO electrode
Goyal RN, et al.
Journal of Pharmaceutical and Biomedical Analysis, 44(5), 1147-1153 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service