Skip to Content
Merck
All Photos(2)

Key Documents

RAB0274

Sigma-Aldrich

Mouse IL-1 β ELISA Kit

for serum, plasma and cell culture supernatant

Synonym(s):

Il-1 beta, Interleukin-1 beta

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32
Pricing and availability is not currently available.

species reactivity

mouse

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type plasma
sample type serum
sample type cell culture supernatant(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 5 pg/mL
standard curve range: 2.74-2000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Il1b(16176)

General description

The Mouse IL-1β ELISA (enzyme-linked immunosorbent assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of mouse IL-1β in serum, plasma, cell culture supernatants and urine. Interleukin-1β (IL-1β) cytokine is generated by monocytes, dendritic cells, macrophages and keratinocytes. Pro-IL-1β, an IL-1β precursor undergoes proteolytic cleavage to produce its active form of 17 kDa.[1] IL-1β is a potent inflammatory cytokine,[1] and mediates inflammation through proinflammatory molecules: cyclooxygenase-2 (COX2) and nitric oxide.[2] IL-1β release is stimulated by injury and increased levels are observed in response to severe pain.[3] It has its main function in T-cell activation, antigen recognition and T-helper cell 17 cell development. IL-1β is also responsible for the formation of IL-17 and IL-22 by nature killer T (NKT) cells and NK cells. It is known to induce inflammation in psoriasis and rheumatoid arthritis.[1] IL-1β controls intercellular adhesion molecule-1 expression. It is significantly associated with bone damage and vascular injury.[4]

Immunogen

Recombinant Mouse IL-1 β

Application

Mouse IL-1 β ELISA Kit has been used to determine the presence of interleukin-1β (IL-1β) in plasma sample.[5] It has also been used to determine the serum concentration levels of IL-1β.[6][7]

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification of interleukin-1 beta as a key mediator in the upregulation of Cav3. 2--USP5 interactions in the pain pathway
Stemkowski PL, et al.
Molecular Pain, 13(36) (2017)
Effect of HI-6 on cytokines production after immunity stimulation by keyhole limpet hemocyanin in a mouse model
Pohanka M
Neuro Endocrinology Letters, 35(4), 101-103 (2014)
Protective effect and mechanism of ginsenoside Rg1 in cerebral ischaemia-reperfusion injury in mice
Wang L, et al
Biomedicine and Pharmacotherapy, 99(4), 876-882 (2018)
Interleukin-1beta induces intercellular adhesion molecule-1 expression, thus enhancing the adhesion between mesenchymal stem cells and endothelial progenitor cells via the p38 MAPK signaling pathway
Guo J, et al.
International Journal of Molecular Medicine, 41(4), 1976-1982 (2018)
Minjie Ju et al.
Molecular medicine reports, 20(4), 3347-3354 (2019-08-23)
Ulinastatin, a urinary trypsin inhibitor (UTI) is commonly used to treat patients with acute inflammatory disease. However, the underlying mechanisms of its anti‑inflammatory effect in acute lung injury (ALI) are not fully understood. The present study aimed to investigate the protective

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service