Skip to Content
Merck
All Photos(1)

Key Documents

SRP3083

Sigma-Aldrich

IL-1 beta human

Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

Catabolin, Endogenous Pyrogen (EP),, Leukocyte Endogenous Mediator (LEM),, Lymphocyte-activating factor (LAF), Mononuclear Cell Factor (MCF)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

≤0.001 ng/mL ED50

mol wt

17.3 kDa

packaging

pkg of 10 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IL1B(3553)

General description

IL-1β (interleukin 1 β) is a proinflammatory cytokine produced in a variety of cells including monocytes, tissue macrophages, keratinocytes and other epithelial cells. It is synthesized as a pro-IL-1β which is changed to active form IL-1β with the help of pro-inflammatory protease caspase-1. IL-1β belongs to the IL-1 gene family. The gene is mapped to human chromosome 2q14. Recombinant human IL-1β is a 17.3kDa protein containing 153 amino acid residues.

Application

IL-1β (interleukin 1 β) human has been used to study the effect of pro- and anti-inflammatory mediators on leukotriene B4 and C4 synthesis as well as secretion in an inflamed porcine endometrium. It has been used to study the effect of cytokines on synthesis and secretion of leukotrienes from endometrial epithelial cells of pigs.

Biochem/physiol Actions

Both IL-1α (interleukin 1 α) and IL-1β binds to the same receptor and has similar if not identical biological properties. These cytokines have a broad range of activities including, stimulation of thymocyte proliferation, by inducing IL-2 release, B-cell maturation and proliferation, mitogenic FGF (fibroblast growth factor)-like activity and the ability to stimulate the release of prostaglandin and collagenase from synovial cells. However, whereas IL-1β is a secreted cytokine, IL-1α is predominantly a cell-associated cytokine. IL-1β is involved in proinflammatory and immunological responses. Some of them include enhancing IL-2 generation, and increasing circulating NO (nitric oxide) and bringing neutrophils. It also regulates gene expression of inflammatory cytokines, mesenchymal growth factors and colony stimulating factors. IL-1β is present in chronic inflamed tissues and also in tissues going for fibrogenesis. It brings neutrophil and macrophage to the site of infection. Mutations in IL-1β gene are linked with renal manifestations and renal sequelae in Henoch-Schönlein purpura. It is also associated with destruction of β-cell in type 1 diabetes.

Sequence

APVRSLNCTL RDSQQKSLVM SGPYELKALH LQGQDMEQQV VFSMSFVQGE ESNDKIPVAL GLKEKNLYLS CVLKDDKPTL QLESVDPKNY PKKKMEKRFV FNKIEINNKL EFESAQFPNW YISTSQAENM PVFLGGTKGG QDITDFTMQF VSS

Physical form

Lyophilized from 10 mM Sodium Phosphate, pH 7.4 + 50mM NaCl.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a stabilizer (example 5% Trehalose) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

ATP is released by monocytes stimulated with pathogen-sensing receptor ligands and induces IL-1beta and IL-18 secretion in an autocrine way.
Piccini A
Proceedings of the National Academy of Sciences of the USA, 105, 8067-8072 (2008)
Xiaohui Guo et al.
Bioscience reports, 40(7) (2020-07-23)
The purpose of the present study was to investigate the specific downstream signaling pathway mediated by PI3K/Akt in resveratrol (RES) anti-apoptosis of nucleus pulposus cells (NPCs). Human NPCs were cultured and divided into six groups. Interleukin (IL)-1β was used to
Proinflammatory cytokine IL-1 beta promotes tumor growth of Lewis lung carcinoma by induction of angiogenic factors: in vivo analysis of tumor-stromal interaction.
Saijo Y
Journal of Immunology, 169, 469-475 (2002)
Pituitary cytokine and growth factor expression and action.
Ray D and Melmed S
Endocrine Reviews, 18, 206-228 (1997)
IL-1 beta induces COX2, MMP-1, -3 and -13, ADAMTS-4, IL-1 beta and IL-6 in human tendon cells.
Tsuzaki M
Journal of Orthopaedic Research : Official Publication of the Orthopaedic Research Society, 21, 256-264 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service