RAB0255
Mouse IL-12 P40/70 ELISA Kit
for serum, plasma and cell culture supernatant
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
species reactivity
mouse
technique(s)
ELISA: suitable
capture ELISA: suitable
shipped in
wet ice
storage temp.
−20°C
Gene Information
mouse ... IL12a(16159) , IL12b(16160)
General description
The Mouse IL-12 (P40/P70) ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of Mouse IL-12 (P40/P70) in serum, plasma, cell culture supernatants and urine.
Immunogen
Recombinant Mouse IL-12 p40 & p70
Application
For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Other Notes
A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.
Please type the word sample in the text box provided for lot number.
Kit Components Also Available Separately
Product No.
Description
SDS
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Acta physiologica (Oxford, England), 212(2), 175-187 (2014-05-09)
The aim of this study was to examine if erythropoietin (EPO) has the potential to act as a biological antioxidant and determine the underlying mechanisms. The rate at which its recombinant form (rHuEPO) reacts with hydroxyl (HO˙), 2,2-diphenyl-1-picrylhydrazyl (DPPH˙) and
International journal of molecular sciences, 15(8), 13938-13966 (2014-08-15)
One of the hallmarks of the acute toxicity of 2,3,7,8-tetrachlorodibenzo-p-dioxin (TCDD) is a drastically reduced feed intake by an unknown mechanism. To further elucidate this wasting syndrome, we followed the effects of a single large dose (100 μg/kg) of TCDD
Theriogenology, 82(3), 427-432 (2014-06-17)
The objective of this study was to compare the concentrations of inflammatory cytokines in uterine flush and serum from healthy postpartum dairy cows and cows with clinical or subclinical endometritis. Clinical endometritis was diagnosed by observation of vaginal discharges (>50%
The Journal of investigative dermatology, 134(11), 2784-2794 (2014-06-18)
A major barrier to successful cancer immunotherapy is the tumor's ability to induce T-cell tolerance by exploiting host regulatory mechanisms. Having discovered the DC-HIL receptor, which inhibits T-cell responses by binding to syndecan-4 on effector T cells, we posited the
The American journal of pathology, 184(11), 2855-2861 (2014-09-02)
Activated microglia are associated with amyloid plaques in transgenic mouse models of cerebral amyloidosis and in human Alzheimer disease; yet, their implication in Alzheimer disease pathogenesis remains unclear. It has been suggested that microglia play dual roles depending on the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service