Skip to Content
Merck
All Photos(2)

Key Documents

RAB0308

Sigma-Aldrich

Mouse IL-6 ELISA Kit

for serum, plasma and cell culture supernatant

Synonym(s):

B-cell stimulatory factor 2, CTL differentiation factor, Hybridoma growth factor, IFN-beta-2, IL-6, Interferon beta-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.84
Pricing and availability is not currently available.

species reactivity

mouse

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type plasma
sample type cell culture supernatant(s)
sample type serum

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 2 pg/mL
standard curve range: 0.82-600 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Il6(24498)

General description

The Mouse IL-6 ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of Mouse IL-6 in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Mouse IL-6

Application

For research use only. Not for use in diagnostic procedures.
Mouse IL-6 ELISA kit has been used for determining the levels of IL-6 in biological samples.[1][2][3]

Biochem/physiol Actions

IL-6 is a pleiotropic cytokine that plays an important role in host defense by regulating immune and inflammatory responses. Produced by T cells, monocytes, fibroblasts, endothelial cells and keratinocytes, IL-6 has diverse biological functions. It stimulates B-cell differentiation and antibody production, synergizes with IL-3 in megakaryocyte development and platelet production, induces expression of hepatic acute-phase proteins, and regulates bone metabolism.[4]

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of HI-6 on cytokines production after immunity stimulation by keyhole limpet hemocyanin in a mouse model.
Pohanka M
Neuro Endocrinology Letters, 35, 155-157 (2014)
The biology of interleukin-6.
Kishimoto T
Blood, 74(1), 1-10 (1989)
Cytocompatible Anti-microbial Dressings of Syzygium cumini Cellulose Nanocrystals Decorated with Silver Nanoparticles Accelerate Acute and Diabetic Wound Healing.
Singla R
Scientific Reports, 7(1), 10457-10457 (2017)
Rubbel Singla et al.
Scientific reports, 7(1), 10457-10457 (2017-09-07)
The ever increasing incidences of non-healing skin wounds have paved way for many efforts on the convoluted process of wound healing. Unfortunately, the lack of relevance and success of modern wound dressings in healing of acute and diabetic wounds still
Laura N Castellani et al.
Scientific reports, 8(1), 772-772 (2018-01-18)
Olanzapine is a widely prescribed antipsychotic drug. While effective in reducing psychoses, treatment with olanzapine causes rapid increases in blood glucose. We wanted to determine if a single bout of exercise, immediately prior to treatment, would attenuate the olanzapine-induced rise

Questions

1–2 of 2 Questions  
  1. Hello. How much should I dilute tissue culture media for the assay? 2-fold? Thanks.

    1 answer
    1. The Assay/Sample Diluent Buffer (Item E2) should be diluted 5-fold. This 1X Assay/Sample Diluent Buffer is used to dilute serum, plasma, or medium samples a recommended 2 fold. Please note that the levels of IL-6 may vary between different samples. Optimal dilution factors for each sample must be determined by the investigator.

      Helpful?

  2. マウスのパイエル板をホモジナイズした上清で測定は可能ですか?

    1 answer
    1. This product is suitable with serum, plasma, and cell culture supernatant. However, it has not been determined if this product can measure the supernatant obtained by homogenizing mouse Peyer's patches. This application will need to be validated by the end-user.

      While the requested ELISA kit nor any of the offered normal ELISA kits are neither designed nor validated with cell/tissue lysate samples, our kits are very accommodating to many different sample types so lysates will most likely work. If it is decided to test these kits with cell/tissue lysate samples, it is recommended to dilute the samples at least 5-fold with Assay Diluent to minimize any effects of the detergents in the lysis buffer. The samples may need to be diluted further but this would need to be determined by the experimenter empirically.

      So for the original lysate, in general it is recommended to shoot for at least 1 mg/ml protein, preferably more, to achieve 50-500 ug/ml after dilution. Again though, this range should just be used as a starting point and the optimal concentration would need to be determined empirically.

      Below are some general guidelines for doing so along with some basic tips for sample preparation.
      Tips on Sample preparation
      In brief, a lysis buffer must meet the following specifications:
      A) has relatively low salt content (700 mM or less)
      B) does not contain sodium azide
      C) does not contain >0.1% SDS
      D) does not contain >10 mM reducing agents (beta-mercaptoethanol or dithiothreitol)
      This would include any buffers used for immunoprecipitations, including RIPA buffer.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service