Skip to Content
Merck
All Photos(1)

Key Documents

MWGF200

Sigma-Aldrich

Gel Filtration Markers Kit for Protein Molecular Weights 12,000-200,000 Da

Synonym(s):

Gel Filtration Protein Ladder

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.25

form

solid

technique(s)

liquid chromatography (LC): suitable

storage temp.

−20°C

General description

Gel filtration markers kit contains five vials of proteins and 1 vial of blue dextran for calibration.

Application

Gel filtration markers are used in protein chromatography and gel filtration chromatography. Gel filtration markers have been used to study glucocorticoid-induced lymphocytolysis as a model system for apoptosis within the immune system. Gel filtration markers have also been used to isolate Polygonatum odoratum lectin, which showed remarkable anti-HSV-II activity towards Vero cells, cytotoxicity towards human melanoma A375 cells and induced apoptosis in a caspase-dependent manner.

Kit Components Also Available Separately

Product No.
Description
SDS

  • A8531Albumin, bovine serum, 66,000 Da MW 1 mL/vialSDS

  • A8656Alcohol Dehydrogenase, yeast, 150,000 Da MW 1 mL/vialSDS

  • A8781β-Amylase, sweet potato, 200,000 Da MW 1 mL/vialSDS

  • D4772 Blue dextran, 2,000,000 Da MW 1 mL/vialSDS

  • C7025Carbonic Anhydrase, bovine erythrocytes 29,000 Da MW 1 mL/vialSDS

  • C7150Cytochrome c, horse heart, 12,400 Da MW 1 mL/vialSDS

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daisuke Seo et al.
Biochimica et biophysica acta. Bioenergetics, 1861(3), 148140-148140 (2019-12-16)
Among the thioredoxin reductase-type ferredoxin-NAD(P)+ oxidoreductase (FNR) family, FNR from photosynthetic purple non‑sulfur bacterium Rhodopseudomonas palustris (RpFNR) is distinctive because the predicted residue on the re-face of the isoalloxazine ring portion of the FAD prosthetic group is a tyrosine. Here
Cristina L Marolda et al.
Journal of bacteriology, 190(6), 2128-2137 (2008-01-22)
Wzz is a membrane protein that determines the chain length distribution of the O-antigen lipopolysaccharide by an unknown mechanism. Wzz proteins consist of two transmembrane helices separated by a large periplasmic loop. The periplasmic loop of Escherichia coli K-12 Wzz
Andreas Krug et al.
The Journal of biological chemistry, 280(1), 585-595 (2004-10-21)
In Corynebacterium glutamicum, the activity of aconitase is 2.5-4-fold higher on propionate, citrate, or acetate than on glucose. Here we show that this variation is caused by transcriptional regulation. In search for putative regulators, a gene (acnR) encoding a TetR-type
Ulrich Strych et al.
BMC microbiology, 7, 40-40 (2007-05-19)
Over the past fifteen years, antibiotic resistance in the Gram-positive opportunistic human pathogen Streptococcus pneumoniae has significantly increased. Clinical isolates from patients with community-acquired pneumonia or otitis media often display resistance to two or more antibiotics. Given the need for
Michael Bussmann et al.
The Journal of biological chemistry, 285(38), 29305-29318 (2010-07-21)
The cg1324 gene (rosR) of Corynebacterium glutamicum encodes a MarR-type transcriptional regulator. By a comparative transcriptome analysis with DNA microarrays of a ΔrosR mutant and the wild type and subsequent EMSAs with purified RosR protein, direct target genes of RosR

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service