Skip to Content
Merck
All Photos(1)

Documents

F1804

Sigma-Aldrich

Monoclonal ANTI-FLAG® M2 antibody produced in mouse

1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)

Synonym(s):

flag antibody, Anti-ddddk, Anti-dykddddk

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.32

biological source

mouse

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

M2, monoclonal

form

buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)

purified by

affinity chromatography (affinity tag purification)

species reactivity

all

concentration

1 mg/mL

isotype

IgG1

immunogen sequence

DYKDDDDK

shipped in

wet ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

The ANTI-FLAG M2 mouse, affinity purified monoclonal antibody binds to fusion proteins containing a FLAG peptide sequence.The antibody recognizes the FLAG peptide sequence at the N-terminus, Met-N-terminus,C-terminus, and internal sites of the fusion protein.

Method of purification - Affinity tag purification

Specificity

Binding site:N-Asp-Tyr-Lys-Asp-Asp-Asp-Asp-Lys-C

Immunogen

FLAG; peptide sequence DYKDDDDK

Application

For highly sensitive and specific detection of FLAG fusion proteins by immunoblotting, immunoprecipitation (IP) , immunohistochemisty, immunofluorescence and immunocyotchemistry. Optimized for single banded detection of FLAG fusion proteins in mammalian, plant, and bacterial expression systems.

Western Blotting and EIA

Learn more product details in our FLAG® application portal.

Physical form

Contains 50% glycerol in phosphate buffered saline.

Preparation Note

Immediately before use, dilute in Tris buffered saline (TBS), pH 8.0, with 3% nonfat Milk

Legal Information

ANTI-FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany
FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

HSV-2 glycoprotein gD targets the CC domain of tetherin and promotes tetherin degradation via lysosomal pathway
Yalan Liu
Virology, 154-154 (2016)
Monika Srivastava et al.
Nature communications, 6, 6253-6253 (2015-02-24)
Roquin is an RNA-binding protein that prevents autoimmunity and inflammation via repression of bound target mRNAs such as inducible costimulator (Icos). When Roquin is absent or mutated (Roquin(san)), Icos is overexpressed in T cells. Here we show that Roquin enhances
ADAR2 regulates RNA stability by modifying access of decay-promoting RNA-binding proteins
Aparna A
Nucleic Acids Research, 45(7), 4189-4201 (2017)
Histone H1 Limits DNA Methylation in Neurospora crassa
Michael Seymour
G3 (Bethesda, Md.) (2016)
Yumin Qiu et al.
Biochimica et biophysica acta. Molecular cell research, 1866(9), 1412-1420 (2019-06-10)
Src is a known proto-oncogene and its aberrant activity is involved in a variety of cancers, including ovarian cancer, whereas the regulatory mechanism of Src has not been fully clarified. In this study, we identified tripartite motif-containing (TRIM) 50 as

Articles

Immunoblotting (Western blot transfer) is a common technique in modern proteomics research.

Related Content

Protein and nucleic acid interaction reagents and resources for investing protein-RNA, protein-DNA, and protein-protein interactions and associated applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service