Skip to Content
Merck
All Photos(1)

Key Documents

D4772

Sigma-Aldrich

Blue Dextran

BioReagent

Synonym(s):

Dextran blue from Leuconostoc mesenteroides

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.25

product line

BioReagent

Quality Level

form

powder

mol wt

~2,000,000

packaging

vial of 50 mg

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Blue dextran is a derivative of dextran and a high molecular weight glucose polymer.

Application

Blue Dextran has been used as a single-phase fluid dye in the mass flow capability of hollow microneedle (HMN). It has also been used as a standard in gel filtration chromatography.

Biochem/physiol Actions

Blue dextran is used in affinity chromatography, gel filtration chromatography, protein chromatography and molecular weight markers. Blue dextran has been used to study Alzheimer′s disease.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and characterization of blue dextrans
Viet D, et al.
Carbohydrate Polymers, 74(3), 372-378 (2008)
Risa Nobuta et al.
Nucleic acids research, 48(18), 10441-10455 (2020-09-18)
Comprehensive genome-wide analysis has revealed the presence of translational elements in the 3' untranslated regions (UTRs) of human transcripts. However, the mechanisms by which translation is initiated in 3' UTRs and the physiological function of their products remain unclear. This
Raja Ghosh et al.
Journal of chromatography. A, 1618, 460892-460892 (2020-01-30)
We discuss how the efficiency of a chromatography device could be enhanced by incorporating a new feature which ensures flow uniformity. The overall flow of fluid within the device, which has a cuboid shape, could be visualized as a combination
Halldor Felde Berg et al.
Reproduction in domestic animals = Zuchthygiene, 55(3), 401-404 (2020-01-10)
An extended lifespan of spermatozoa following artificial insemination (AI) can make the timing of insemination less critical, as previously demonstrated with immobilized spermatozoa that are gradually released from an alginate gel. The purpose was to examine the in vivo dissolution
D L Roerig et al.
The American journal of physiology, 262(3 Pt 2), H728-H733 (1992-03-01)
We investigated the uptake and efflux of Blue Dextran in the isolated perfused rabbit lung. Blue Dextran is a high-molecular-weight glucose polymer (original mol wt 2 x 10(6) g/mol) containing covalently bonded Reactive Blue 2 dye (approximately mmol/g dextran). This

Protocols

Gel filtration chromatography is an established method for determining the size and molecular mass of proteins.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service