Skip to Content
Merck
All Photos(1)

Key Documents

GE17-0445-01

Calibration Kit High Molecular Weight For Electrophoresis

Cytiva 17-0445-01, pkg of 10 vials

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105323
NACRES:
NA.32

form

solid

feature

wetted part: no

packaging

pkg of 10 vials

manufacturer/tradename

Cytiva 17-0445-01

storage condition

(Unopened vials should be s-red at 4°C)

color

White

storage temp.

2-8°C

General description

The calibration kit is specifically designed for electrophoresis applications, providing precise standards for accurate molecular weight determination.

Features and Benefits

  • Determine molecular weight of uncharacterized Protein samples with precision by comparison with carefully selected standards.
  • Get clear, sharp bands for easy measurement of migration distances.
  • Bands can be visualized by using Deep Purple Total Protein Stain, Coomassie Brilliant Blue, or silver staining.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service