Skip to Content
Merck
All Photos(4)

Key Documents

M5375

Sigma-Aldrich

D-(+)-Melezitose hydrate

≥97% (HPLC)

Synonym(s):

α-D-Glc-[1→3]-β-D-Fru-[2→1]-α-D-Glc

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H32O16 · xH2O
CAS Number:
Molecular Weight:
504.44 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

honey bee (honey)

Assay

≥97% (HPLC)

form

powder

optical activity

[α]/D 86.8 to 89.2 °, c = 4% (w/v) in water

color

white

mp

160 °C (dec.) (lit.)

solubility

water: 50 mg/mL, clear, colorless

storage temp.

room temp

SMILES string

[H]O[H].OC[C@H]1O[C@H](O[C@H]2[C@H](O)[C@@H](CO)O[C@@]2(CO)O[C@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C18H32O16.H2O/c19-1-5-8(23)11(26)13(28)16(30-5)32-15-10(25)7(3-21)33-18(15,4-22)34-17-14(29)12(27)9(24)6(2-20)31-17;/h5-17,19-29H,1-4H2;1H2/t5-,6-,7-,8-,9-,10-,11+,12+,13-,14-,15+,16-,17-,18+;/m1./s1

InChI key

CFXAVQUXSYFPDE-ZDVKUAOASA-N

Looking for similar products? Visit Product Comparison Guide

General description

D-Melezitose is a carbon substrate.

Application

D-Melezitose has been used in a study to assess the identification of clinical isolates of indole-positive and indole-negative Klebsiella spp. It has also been used in a study to investigate new epidemiologic markers of Klebsiella oxytoca strains.

Other Notes

To gain a comprehensive understanding of our extensive range of Oligosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Richard et al.
Annales de biologie clinique, 45(4), 397-401 (1987-01-01)
In addition to capsular antigen typing and biotyping (sorbose, d-tartrate, dulcitol), the epidemiology of Klebsiella oxytoca can be improved by new biochemical characterization. The authors propose to determine tetrathionate-reductase activity, D-melezitose fermentation and brown pigment production onto a mineral gluconate-ferric
D Monnet et al.
Journal of clinical microbiology, 32(4), 1121-1122 (1994-04-01)
Conventional methods usually fail to identify Klebsiella planticola and Klebsiella terrigena, which represent up to 19% of clinical Klebsiella isolates. By combining four carbon substrate assimilation tests and two conventional tests, the method identified these species with a specificity and
Maria Silvana Alves et al.
Journal of clinical microbiology, 44(10), 3640-3646 (2006-08-25)
Biochemical methods employed to classify bacterial species have limitations and may have contributed to the taxonomic complexity recently reported for the genus Klebsiella. The objective of the present study was to apply a simple biochemical test panel to classify a
Isabel González-Delgado et al.
Journal of agricultural and food chemistry, 65(8), 1649-1658 (2017-02-09)
A rational optimization for the synthesis of galacto-oligosaccharides (GOS) from lactose catalyzed by β-galactosidase from Aspergillus aculeatus, included in the commercial product Pectinex Ultra SP-L, has been performed by using experimental design and surface response methodology. This accurate tool optimized
Robert W P Glowacki et al.
Cell host & microbe, 27(1), 79-92 (2020-01-07)
Efficient nutrient acquisition in the human gut is essential for microbial persistence. Although polysaccharides have been well-studied nutrients for the gut microbiome, other resources such as nucleic acids and nucleosides are less studied. We describe several ribose-utilization systems (RUSs) that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service